Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84221958-84222219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIT2
Entrez Gene ID
340745
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 2
Gene Aliases
LRRC22
UniGene
Hs.450425
Gene Chromosome Location
Chr.10: 84220485-84225589 on Build GRCh38
Transcripts
NM_001017924.3 NM_001284223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4807996 c.1512G>A p.R504R Chr.10: 84222061-84222061 on Build GRCh38
6048397 c.1528delA p.T510fs*26 Chr.10: 84222045-84222045 on Build GRCh38
685186 c.1494C>T p.R498R Chr.10: 84222079-84222079 on Build GRCh38
6048398 c.1558delA p.T520fs*26 Chr.10: 84222045-84222045 on Build GRCh38
6310446 c.1558A>C p.T520P Chr.10: 84222045-84222045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74145770 Chr.10: 84222000-84222000 on Build GRCh38
rs769319630 Chr.10: 84222145-84222145 on Build GRCh38
rs767381061 Chr.10: 84222170-84222170 on Build GRCh38
rs781380404 Chr.10: 84222134-84222134 on Build GRCh38
rs757386251 Chr.10: 84222117-84222117 on Build GRCh38
rs761320330 Chr.10: 84222004-84222004 on Build GRCh38
rs766893500 Chr.10: 84222189-84222189 on Build GRCh38
rs758570988 Chr.10: 84222072-84222072 on Build GRCh38
rs148642694 Chr.10: 84222162-84222162 on Build GRCh38
rs752102061 Chr.10: 84222190-84222190 on Build GRCh38
rs759431898 Chr.10: 84222165-84222165 on Build GRCh38
rs543135655 Chr.10: 84222131-84222131 on Build GRCh38
rs779422015 Chr.10: 84221982-84221982 on Build GRCh38
rs140185624 Chr.10: 84222079-84222079 on Build GRCh38
rs374527427 Chr.10: 84221983-84221983 on Build GRCh38
rs781664775 Chr.10: 84222195-84222195 on Build GRCh38
rs941827293 Chr.10: 84221999-84221999 on Build GRCh38
rs375340247 Chr.10: 84222109-84222109 on Build GRCh38
rs772301486 Chr.10: 84222104-84222105 on Build GRCh38
rs868157068 Chr.10: 84222128-84222128 on Build GRCh38
rs6585847 Chr.10: 84222045-84222045 on Build GRCh38
rs757309860 Chr.10: 84222034-84222034 on Build GRCh38
rs146624367 Chr.10: 84222006-84222006 on Build GRCh38
rs12217769 Chr.10: 84222086-84222086 on Build GRCh38
rs747387009 Chr.10: 84222132-84222132 on Build GRCh38
rs370294340 Chr.10: 84222119-84222119 on Build GRCh38
rs762954857 Chr.10: 84222011-84222011 on Build GRCh38
rs372043491 Chr.10: 84222081-84222081 on Build GRCh38
rs772847925 Chr.10: 84222156-84222156 on Build GRCh38
rs559063239 Chr.10: 84221994-84221994 on Build GRCh38
rs753881269 Chr.10: 84222032-84222032 on Build GRCh38
rs760447728 Chr.10: 84222157-84222157 on Build GRCh38
rs776287901 Chr.10: 84222066-84222066 on Build GRCh38
rs765371341 Chr.10: 84222179-84222179 on Build GRCh38
rs142100979 Chr.10: 84222181-84222181 on Build GRCh38
rs143323013 Chr.10: 84222056-84222056 on Build GRCh38
rs777236097 Chr.10: 84222198-84222198 on Build GRCh38
rs916361797 Chr.10: 84222172-84222172 on Build GRCh38
rs907196641 Chr.10: 84222031-84222031 on Build GRCh38
rs775798595 Chr.10: 84221987-84221987 on Build GRCh38
rs747366884 Chr.10: 84222063-84222063 on Build GRCh38
rs147546091 Chr.10: 84222065-84222065 on Build GRCh38
rs747885471 Chr.10: 84222138-84222138 on Build GRCh38
rs774295334 Chr.10: 84222094-84222094 on Build GRCh38
rs755798375 Chr.10: 84222197-84222197 on Build GRCh38
rs567372398 Chr.10: 84222150-84222150 on Build GRCh38
rs150620602 Chr.10: 84222042-84222042 on Build GRCh38
rs776071626 Chr.10: 84222003-84222003 on Build GRCh38
rs891325702 Chr.10: 84222153-84222153 on Build GRCh38
rs546980718 Chr.10: 84222178-84222178 on Build GRCh38
rs190259697 Chr.10: 84222111-84222111 on Build GRCh38
rs746859849 Chr.10: 84222131-84222132 on Build GRCh38
rs1011538149 Chr.10: 84222200-84222200 on Build GRCh38
rs759350344 Chr.10: 84222108-84222108 on Build GRCh38
rs553741973 Chr.10: 84222021-84222021 on Build GRCh38
rs778965402 Chr.10: 84222032-84222032 on Build GRCh38
rs771009302 Chr.10: 84222158-84222158 on Build GRCh38
rs759139373 Chr.10: 84222105-84222106 on Build GRCh38
rs764248986 Chr.10: 84222110-84222110 on Build GRCh38
rs891987741 Chr.10: 84222040-84222040 on Build GRCh38
rs146667369 Chr.10: 84222187-84222187 on Build GRCh38
rs750486199 Chr.10: 84222124-84222124 on Build GRCh38
rs755418464 Chr.10: 84222191-84222191 on Build GRCh38
rs527413911 Chr.10: 84222008-84222008 on Build GRCh38
rs866194422 Chr.10: 84222069-84222069 on Build GRCh38
rs531991237 Chr.10: 84222177-84222177 on Build GRCh38
rs766327815 Chr.10: 84222017-84222017 on Build GRCh38
rs765404571 Chr.10: 84222120-84222120 on Build GRCh38
rs758969467 Chr.10: 84222125-84222125 on Build GRCh38
rs947932172 Chr.10: 84222014-84222014 on Build GRCh38
rs766565567 Chr.10: 84222080-84222080 on Build GRCh38
rs748327683 Chr.10: 84222051-84222052 on Build GRCh38
rs771351258 Chr.10: 84221979-84221979 on Build GRCh38
rs12218853 Chr.10: 84222133-84222133 on Build GRCh38
rs778079567 Chr.10: 84222029-84222029 on Build GRCh38
rs756473406 Chr.10: 84222022-84222022 on Build GRCh38

View Full Product Details