Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40681077-40681330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCHR1
Entrez Gene ID
2847
Gene Name
melanin concentrating hormone receptor 1
Gene Aliases
GPR24, MCH-1R, MCH1R, SLC-1, SLC1
UniGene
Hs.248122
Gene Chromosome Location
Chr.22: 40679178-40682814 on Build GRCh38
Transcripts
NM_005297.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3554851 c.540T>A p.F180L Chr.22: 40681199-40681199 on Build GRCh38
4701138 c.600C>A p.T200T Chr.22: 40681259-40681259 on Build GRCh38
1034480 c.578C>T p.A193V Chr.22: 40681237-40681237 on Build GRCh38
4618203 c.492C>A p.G164G Chr.22: 40681151-40681151 on Build GRCh38
1034481 c.663G>T p.T221T Chr.22: 40681322-40681322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200659421 Chr.22: 40681161-40681161 on Build GRCh38
rs751739288 Chr.22: 40681269-40681269 on Build GRCh38
rs915365004 Chr.22: 40681237-40681237 on Build GRCh38
rs1007338393 Chr.22: 40681128-40681128 on Build GRCh38
rs201977776 Chr.22: 40681231-40681231 on Build GRCh38
rs762273865 Chr.22: 40681105-40681105 on Build GRCh38
rs201299757 Chr.22: 40681252-40681252 on Build GRCh38
rs375716449 Chr.22: 40681187-40681187 on Build GRCh38
rs770353095 Chr.22: 40681233-40681233 on Build GRCh38
rs752123648 Chr.22: 40681124-40681124 on Build GRCh38
rs757056931 Chr.22: 40681173-40681173 on Build GRCh38
rs746908607 Chr.22: 40681308-40681308 on Build GRCh38
rs745882964 Chr.22: 40681178-40681178 on Build GRCh38
rs948134494 Chr.22: 40681293-40681293 on Build GRCh38
rs760177693 Chr.22: 40681265-40681265 on Build GRCh38
rs80015181 Chr.22: 40681269-40681269 on Build GRCh38
rs763368168 Chr.22: 40681119-40681119 on Build GRCh38
rs773420850 Chr.22: 40681243-40681243 on Build GRCh38
rs776954401 Chr.22: 40681101-40681101 on Build GRCh38
rs748371706 Chr.22: 40681232-40681232 on Build GRCh38
rs200107865 Chr.22: 40681190-40681190 on Build GRCh38
rs764292808 Chr.22: 40681189-40681209 on Build GRCh38
rs987667577 Chr.22: 40681148-40681148 on Build GRCh38
rs758174382 Chr.22: 40681297-40681297 on Build GRCh38
rs180949089 Chr.22: 40681309-40681309 on Build GRCh38
rs368842613 Chr.22: 40681216-40681216 on Build GRCh38
rs199740916 Chr.22: 40681278-40681278 on Build GRCh38
rs750210146 Chr.22: 40681288-40681288 on Build GRCh38
rs200659135 Chr.22: 40681146-40681146 on Build GRCh38
rs775425755 Chr.22: 40681095-40681095 on Build GRCh38
rs979674247 Chr.22: 40681226-40681226 on Build GRCh38
rs778664994 Chr.22: 40681176-40681176 on Build GRCh38
rs200876813 Chr.22: 40681218-40681218 on Build GRCh38
rs757274861 Chr.22: 40681222-40681222 on Build GRCh38
rs761007989 Chr.22: 40681099-40681099 on Build GRCh38
rs200222615 Chr.22: 40681271-40681271 on Build GRCh38
rs779995029 Chr.22: 40681198-40681198 on Build GRCh38
rs765055919 Chr.22: 40681287-40681287 on Build GRCh38
rs188147970 Chr.22: 40681279-40681279 on Build GRCh38
rs201390387 Chr.22: 40681296-40681296 on Build GRCh38
rs199947845 Chr.22: 40681240-40681240 on Build GRCh38
rs200897081 Chr.22: 40681100-40681100 on Build GRCh38
rs202175341 Chr.22: 40681253-40681253 on Build GRCh38
rs766813535 Chr.22: 40681258-40681258 on Build GRCh38
rs774878903 Chr.22: 40681264-40681264 on Build GRCh38
rs146647078 Chr.22: 40681172-40681172 on Build GRCh38
rs201895311 Chr.22: 40681117-40681117 on Build GRCh38
rs574065876 Chr.22: 40681210-40681210 on Build GRCh38
rs779860748 Chr.22: 40681299-40681299 on Build GRCh38
rs369827677 Chr.22: 40681123-40681123 on Build GRCh38
rs199923574 Chr.22: 40681151-40681151 on Build GRCh38
rs140185504 Chr.22: 40681225-40681225 on Build GRCh38
rs763567756 Chr.22: 40681250-40681250 on Build GRCh38
rs560068779 Chr.22: 40681166-40681166 on Build GRCh38

View Full Product Details