Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48742080-48742343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IZUMO1
Entrez Gene ID
284359
Gene Name
izumo sperm-egg fusion 1
Gene Aliases
IZUMO, OBF
UniGene
Hs.400688
Gene Chromosome Location
Chr.19: 48740888-48747499 on Build GRCh38
Transcripts
NM_001321864.1 NM_001321865.1 NM_182575.2 NR_135832.1 XM_005258793.4 XM_011526811.2 XM_011526812.2 XM_011526813.2 XM_011526815.2 XM_017026646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4079906 c.594T>G p.F198L Chr.19: 48742215-48742215 on Build GRCh38
4693699 c.569C>T p.S190L Chr.19: 48742240-48742240 on Build GRCh38
5631854 c.600G>A p.R200R Chr.19: 48742209-48742209 on Build GRCh38
6689169 c.500-3delC p.? Chr.19: 48742312-48742312 on Build GRCh38
7256298 c.575G>T p.G192V Chr.19: 48742234-48742234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770076028 Chr.19: 48742310-48742310 on Build GRCh38
rs369730028 Chr.19: 48742175-48742175 on Build GRCh38
rs769405592 Chr.19: 48742120-48742120 on Build GRCh38
rs748508427 Chr.19: 48742161-48742161 on Build GRCh38
rs770771867 Chr.19: 48742322-48742322 on Build GRCh38
rs770575579 Chr.19: 48742186-48742186 on Build GRCh38
rs534378206 Chr.19: 48742122-48742122 on Build GRCh38
rs759540044 Chr.19: 48742207-48742207 on Build GRCh38
rs762502995 Chr.19: 48742184-48742184 on Build GRCh38
rs765455055 Chr.19: 48742246-48742246 on Build GRCh38
rs750628658 Chr.19: 48742249-48742249 on Build GRCh38
rs559239497 Chr.19: 48742134-48742134 on Build GRCh38
rs760422018 Chr.19: 48742243-48742243 on Build GRCh38
rs140133186 Chr.19: 48742306-48742306 on Build GRCh38
rs755456942 Chr.19: 48742302-48742302 on Build GRCh38
rs767410011 Chr.19: 48742231-48742231 on Build GRCh38
rs575148968 Chr.19: 48742265-48742265 on Build GRCh38
rs542242649 Chr.19: 48742314-48742314 on Build GRCh38
rs775384269 Chr.19: 48742130-48742130 on Build GRCh38
rs908310347 Chr.19: 48742155-48742155 on Build GRCh38
rs372968209 Chr.19: 48742316-48742316 on Build GRCh38
rs1002210285 Chr.19: 48742301-48742301 on Build GRCh38
rs551903814 Chr.19: 48742307-48742307 on Build GRCh38
rs758406565 Chr.19: 48742261-48742261 on Build GRCh38
rs1009994209 Chr.19: 48742313-48742313 on Build GRCh38
rs114817513 Chr.19: 48742202-48742202 on Build GRCh38
rs146107337 Chr.19: 48742240-48742240 on Build GRCh38
rs149283178 Chr.19: 48742239-48742239 on Build GRCh38
rs748505096 Chr.19: 48742308-48742308 on Build GRCh38
rs1032840408 Chr.19: 48742113-48742113 on Build GRCh38
rs577367047 Chr.19: 48742145-48742145 on Build GRCh38
rs1023999698 Chr.19: 48742129-48742129 on Build GRCh38
rs769454388 Chr.19: 48742167-48742167 on Build GRCh38

View Full Product Details