Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38499250-38499519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGAPB
Entrez Gene ID
57148
Gene Name
Ral GTPase activating protein non-catalytic beta subunit
Gene Aliases
KIAA1219, RalGAPbeta
UniGene
Hs.598850
Gene Chromosome Location
Chr.20: 38472806-38578861 on Build GRCh38
Transcripts
NM_001282917.1 NM_001282918.1 NM_020336.3 XM_005260462.2 XM_005260463.2 XM_005260464.2 XM_005260465.2 XM_017027966.1 XM_017027967.1 XM_017027968.1 XM_017027969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752940 c.626G>C p.R209P Chr.20: 38499519-38499519 on Build GRCh38
1026653 c.567G>T p.E189D Chr.20: 38499460-38499460 on Build GRCh38
7097535 c.567G>A p.E189E Chr.20: 38499460-38499460 on Build GRCh38
246944 c.620G>A p.C207Y Chr.20: 38499513-38499513 on Build GRCh38
1495096 c.606G>A p.V202V Chr.20: 38499499-38499499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540535343 Chr.20: 38499493-38499493 on Build GRCh38
rs201059121 Chr.20: 38499425-38499425 on Build GRCh38
rs369928478 Chr.20: 38499418-38499418 on Build GRCh38
rs570998493 Chr.20: 38499367-38499367 on Build GRCh38
rs1051605703 Chr.20: 38499397-38499397 on Build GRCh38
rs775580865 Chr.20: 38499490-38499490 on Build GRCh38
rs563293178 Chr.20: 38499310-38499310 on Build GRCh38
rs780968226 Chr.20: 38499405-38499416 on Build GRCh38
rs914227737 Chr.20: 38499329-38499329 on Build GRCh38
rs772132622 Chr.20: 38499485-38499485 on Build GRCh38
rs201959804 Chr.20: 38499430-38499430 on Build GRCh38
rs745956799 Chr.20: 38499483-38499483 on Build GRCh38
rs199873106 Chr.20: 38499415-38499415 on Build GRCh38
rs757228863 Chr.20: 38499474-38499474 on Build GRCh38
rs766022300 Chr.20: 38499278-38499278 on Build GRCh38
rs947080819 Chr.20: 38499373-38499373 on Build GRCh38
rs777694694 Chr.20: 38499452-38499452 on Build GRCh38
rs974463581 Chr.20: 38499376-38499376 on Build GRCh38
rs145596161 Chr.20: 38499492-38499492 on Build GRCh38
rs992239669 Chr.20: 38499361-38499361 on Build GRCh38
rs753352868 Chr.20: 38499385-38499385 on Build GRCh38
rs760960660 Chr.20: 38499421-38499421 on Build GRCh38
rs746254036 Chr.20: 38499406-38499409 on Build GRCh38
rs749284020 Chr.20: 38499464-38499464 on Build GRCh38
rs16987350 Chr.20: 38499437-38499437 on Build GRCh38
rs921734739 Chr.20: 38499382-38499382 on Build GRCh38
rs754474811 Chr.20: 38499432-38499432 on Build GRCh38
rs774269467 Chr.20: 38499402-38499402 on Build GRCh38
rs764376448 Chr.20: 38499423-38499423 on Build GRCh38
rs943618618 Chr.20: 38499451-38499451 on Build GRCh38
rs370244476 Chr.20: 38499438-38499438 on Build GRCh38
rs778868222 Chr.20: 38499478-38499478 on Build GRCh38

View Full Product Details