Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186312080-186312329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Gene Symbol
TPR
Entrez Gene ID
7175
Gene Name
translocated promoter region, nuclear basket protein
Gene Aliases
-
UniGene
Hs.279640
Gene Chromosome Location
Chr.1: 186311652-186375325 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6345305 c.3847G>T p.G1283* Chr.1: 186312228-186312228 on Build GRCh38
3789244 c.3851G>A p.R1284K Chr.1: 186312232-186312232 on Build GRCh38
6755454 c.3794-7A>C p.? Chr.1: 186312168-186312168 on Build GRCh38
4901144 c.3888G>A p.T1296T Chr.1: 186312269-186312269 on Build GRCh38
4433167 c.3902G>A p.R1301K Chr.1: 186312283-186312283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138853571 Chr.1: 186312302-186312302 on Build GRCh38
rs865966866 Chr.1: 186312138-186312138 on Build GRCh38
rs777069099 Chr.1: 186312304-186312304 on Build GRCh38
rs745600232 Chr.1: 186312160-186312160 on Build GRCh38
rs755088130 Chr.1: 186312126-186312126 on Build GRCh38
rs34326122 Chr.1: 186312148-186312149 on Build GRCh38
rs12134934 Chr.1: 186312268-186312268 on Build GRCh38
rs942356063 Chr.1: 186312245-186312245 on Build GRCh38
rs777908416 Chr.1: 186312157-186312157 on Build GRCh38
rs940640055 Chr.1: 186312128-186312128 on Build GRCh38
rs771197949 Chr.1: 186312297-186312297 on Build GRCh38
rs772710281 Chr.1: 186312178-186312195 on Build GRCh38
rs376436154 Chr.1: 186312129-186312129 on Build GRCh38
rs772720067 Chr.1: 186312294-186312294 on Build GRCh38
rs773912832 Chr.1: 186312187-186312187 on Build GRCh38
rs749492772 Chr.1: 186312159-186312162 on Build GRCh38
rs1043443207 Chr.1: 186312238-186312238 on Build GRCh38
rs369108138 Chr.1: 186312256-186312256 on Build GRCh38
rs755610521 Chr.1: 186312227-186312227 on Build GRCh38
rs774298312 Chr.1: 186312286-186312286 on Build GRCh38
rs747293786 Chr.1: 186312262-186312262 on Build GRCh38
rs747620778 Chr.1: 186312288-186312288 on Build GRCh38
rs918794884 Chr.1: 186312157-186312157 on Build GRCh38
rs369156773 Chr.1: 186312141-186312141 on Build GRCh38
rs768509513 Chr.1: 186312285-186312285 on Build GRCh38
rs771654669 Chr.1: 186312289-186312289 on Build GRCh38
rs565193791 Chr.1: 186312269-186312269 on Build GRCh38
rs910974224 Chr.1: 186312210-186312210 on Build GRCh38
rs747775887 Chr.1: 186312168-186312168 on Build GRCh38
rs373568605 Chr.1: 186312127-186312127 on Build GRCh38
rs375198741 Chr.1: 186312178-186312178 on Build GRCh38
rs543150876 Chr.1: 186312124-186312124 on Build GRCh38
rs747928688 Chr.1: 186312128-186312133 on Build GRCh38
rs779954280 Chr.1: 186312270-186312270 on Build GRCh38
rs907871271 Chr.1: 186312125-186312125 on Build GRCh38
rs902185020 Chr.1: 186312251-186312251 on Build GRCh38
rs749214186 Chr.1: 186312284-186312284 on Build GRCh38
rs769585191 Chr.1: 186312133-186312136 on Build GRCh38
rs760435378 Chr.1: 186312295-186312295 on Build GRCh38
rs754385580 Chr.1: 186312222-186312222 on Build GRCh38
rs771201446 Chr.1: 186312163-186312164 on Build GRCh38
rs760919750 Chr.1: 186312213-186312213 on Build GRCh38
rs769660657 Chr.1: 186312168-186312168 on Build GRCh38
rs138093894 Chr.1: 186312260-186312260 on Build GRCh38
rs778069259 Chr.1: 186312152-186312152 on Build GRCh38
rs1293985 Chr.1: 186312196-186312196 on Build GRCh38
rs766817116 Chr.1: 186312217-186312217 on Build GRCh38
rs759992615 Chr.1: 186312307-186312307 on Build GRCh38
rs879468274 Chr.1: 186312266-186312266 on Build GRCh38
rs759630200 Chr.1: 186312176-186312176 on Build GRCh38
rs767032008 Chr.1: 186312194-186312194 on Build GRCh38
rs749269200 Chr.1: 186312172-186312172 on Build GRCh38
rs774560899 Chr.1: 186312166-186312173 on Build GRCh38
rs765843977 Chr.1: 186312236-186312236 on Build GRCh38
rs761330232 Chr.1: 186312188-186312188 on Build GRCh38

View Full Product Details