Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9552423-9552664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHFPL4
Entrez Gene ID
375323
Gene Name
lipoma HMGIC fusion partner-like 4
Gene Aliases
-
UniGene
Hs.56782
Gene Chromosome Location
Chr.3: 9498361-9553802 on Build GRCh38
Transcripts
NM_198560.2 XM_017006359.1 XM_017006360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644734 c.137G>T p.W46L Chr.3: 9552543-9552543 on Build GRCh38
402285 c.62C>A p.A21D Chr.3: 9552618-9552618 on Build GRCh38
3974604 c.103A>G p.I35V Chr.3: 9552577-9552577 on Build GRCh38
6566790 c.58C>T p.R20W Chr.3: 9552622-9552622 on Build GRCh38
1425783 c.144C>T p.G48G Chr.3: 9552536-9552536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753941704 Chr.3: 9552630-9552630 on Build GRCh38
rs746253248 Chr.3: 9552638-9552638 on Build GRCh38
rs766047581 Chr.3: 9552569-9552569 on Build GRCh38
rs756217548 Chr.3: 9552464-9552464 on Build GRCh38
rs779165564 Chr.3: 9552634-9552634 on Build GRCh38
rs757488524 Chr.3: 9552631-9552631 on Build GRCh38
rs746226883 Chr.3: 9552522-9552522 on Build GRCh38
rs748291845 Chr.3: 9552458-9552458 on Build GRCh38
rs761342548 Chr.3: 9552551-9552551 on Build GRCh38
rs769117293 Chr.3: 9552562-9552562 on Build GRCh38
rs377128905 Chr.3: 9552616-9552616 on Build GRCh38
rs755028674 Chr.3: 9552443-9552443 on Build GRCh38
rs370030672 Chr.3: 9552642-9552642 on Build GRCh38
rs867351413 Chr.3: 9552622-9552622 on Build GRCh38
rs779203728 Chr.3: 9552490-9552490 on Build GRCh38
rs777658749 Chr.3: 9552624-9552624 on Build GRCh38
rs775927253 Chr.3: 9552540-9552540 on Build GRCh38
rs9880840 Chr.3: 9552566-9552566 on Build GRCh38
rs201993485 Chr.3: 9552614-9552614 on Build GRCh38
rs781248542 Chr.3: 9552448-9552448 on Build GRCh38
rs751347672 Chr.3: 9552572-9552572 on Build GRCh38
rs752644447 Chr.3: 9552611-9552611 on Build GRCh38
rs749406469 Chr.3: 9552467-9552467 on Build GRCh38
rs373908621 Chr.3: 9552529-9552529 on Build GRCh38
rs200109698 Chr.3: 9552599-9552599 on Build GRCh38
rs370835564 Chr.3: 9552623-9552623 on Build GRCh38
rs138010331 Chr.3: 9552465-9552465 on Build GRCh38
rs759386978 Chr.3: 9552598-9552598 on Build GRCh38
rs575246923 Chr.3: 9552563-9552563 on Build GRCh38
rs370762541 Chr.3: 9552442-9552442 on Build GRCh38
rs771341752 Chr.3: 9552478-9552478 on Build GRCh38

View Full Product Details