Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 33100462-33100693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RP9
Entrez Gene ID
6100
Gene Name
retinitis pigmentosa 9 (autosomal dominant)
Gene Aliases
PAP-1, PAP1
UniGene
Hs.326805
Gene Chromosome Location
Chr.7: 33094797-33109390 on Build GRCh38
Transcripts
NM_203288.1 XM_011515468.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4722499 c.157G>A p.E53K Chr.7: 33100557-33100557 on Build GRCh38
4752382 c.173G>A p.G58E Chr.7: 33100541-33100541 on Build GRCh38
3832684 c.169C>A p.P57T Chr.7: 33100545-33100545 on Build GRCh38
3228317 c.169C>T p.P57S Chr.7: 33100545-33100545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888456985 Chr.7: 33100497-33100497 on Build GRCh38
rs1056182066 Chr.7: 33100607-33100607 on Build GRCh38
rs759895247 Chr.7: 33100657-33100657 on Build GRCh38
rs768209317 Chr.7: 33100583-33100583 on Build GRCh38
rs775295369 Chr.7: 33100577-33100577 on Build GRCh38
rs765382810 Chr.7: 33100664-33100664 on Build GRCh38
rs531193736 Chr.7: 33100636-33100636 on Build GRCh38
rs769508310 Chr.7: 33100576-33100576 on Build GRCh38
rs7792378 Chr.7: 33100623-33100623 on Build GRCh38
rs746958575 Chr.7: 33100526-33100526 on Build GRCh38
rs753704162 Chr.7: 33100640-33100640 on Build GRCh38
rs776941168 Chr.7: 33100606-33100606 on Build GRCh38
rs368775243 Chr.7: 33100587-33100587 on Build GRCh38
rs745693022 Chr.7: 33100563-33100563 on Build GRCh38
rs758482137 Chr.7: 33100512-33100512 on Build GRCh38
rs115938195 Chr.7: 33100558-33100558 on Build GRCh38
rs943970488 Chr.7: 33100573-33100573 on Build GRCh38
rs761220604 Chr.7: 33100594-33100594 on Build GRCh38
rs371758891 Chr.7: 33100601-33100601 on Build GRCh38
rs770824356 Chr.7: 33100557-33100557 on Build GRCh38
rs985174175 Chr.7: 33100525-33100525 on Build GRCh38
rs552249692 Chr.7: 33100618-33100618 on Build GRCh38
rs777770953 Chr.7: 33100515-33100515 on Build GRCh38
rs762753710 Chr.7: 33100582-33100582 on Build GRCh38

View Full Product Details