Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46989482-46989723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBEAL2
Entrez Gene ID
23218
Gene Name
neurobeachin like 2
Gene Aliases
BDPLT4, GPS
UniGene
Hs.437043
Gene Chromosome Location
Chr.3: 46979683-47009704 on Build GRCh38
Transcripts
NM_015175.2 XM_006713072.3 XM_011533533.2 XM_017006010.1 XM_017006011.1 XM_017006012.1 XM_017006013.1 XM_017006014.1 XM_017006015.1 XM_017006016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6722427 c.527T>C p.L176S Chr.3: 46989564-46989564 on Build GRCh38
5974807 c.531C>T p.P177P Chr.3: 46989568-46989568 on Build GRCh38
4499742 c.549C>T p.F183F Chr.3: 46989586-46989586 on Build GRCh38
6316369 c.478G>T p.V160F Chr.3: 46989515-46989515 on Build GRCh38
3916160 c.513C>T p.F171F Chr.3: 46989550-46989550 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774917873 Chr.3: 46989611-46989611 on Build GRCh38
rs754094112 Chr.3: 46989536-46989536 on Build GRCh38
rs757417910 Chr.3: 46989558-46989558 on Build GRCh38
rs375718269 Chr.3: 46989657-46989657 on Build GRCh38
rs758499927 Chr.3: 46989580-46989580 on Build GRCh38
rs777095694 Chr.3: 46989617-46989617 on Build GRCh38
rs759901172 Chr.3: 46989620-46989621 on Build GRCh38
rs745890949 Chr.3: 46989581-46989581 on Build GRCh38
rs771485869 Chr.3: 46989605-46989616 on Build GRCh38
rs866793907 Chr.3: 46989521-46989521 on Build GRCh38
rs370943621 Chr.3: 46989615-46989615 on Build GRCh38
rs1009437569 Chr.3: 46989585-46989585 on Build GRCh38
rs559872924 Chr.3: 46989653-46989653 on Build GRCh38
rs900966116 Chr.3: 46989610-46989610 on Build GRCh38
rs200476302 Chr.3: 46989565-46989565 on Build GRCh38
rs767271874 Chr.3: 46989527-46989527 on Build GRCh38
rs908294557 Chr.3: 46989663-46989663 on Build GRCh38
rs1005160380 Chr.3: 46989700-46989700 on Build GRCh38
rs1019466579 Chr.3: 46989603-46989603 on Build GRCh38
rs766408127 Chr.3: 46989504-46989504 on Build GRCh38
rs780666612 Chr.3: 46989584-46989584 on Build GRCh38
rs750930405 Chr.3: 46989553-46989553 on Build GRCh38
rs375726193 Chr.3: 46989507-46989507 on Build GRCh38
rs747817191 Chr.3: 46989630-46989630 on Build GRCh38
rs923419221 Chr.3: 46989604-46989604 on Build GRCh38
rs577292787 Chr.3: 46989608-46989608 on Build GRCh38

View Full Product Details