Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18261153-18261426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB2
Entrez Gene ID
783
Gene Name
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases
CACNLB2, CAVB2, MYSB
UniGene
Hs.59093
Gene Chromosome Location
Chr.10: 18140560-18545881 on Build GRCh38
Transcripts
NM_000724.3 NM_001167945.1 NM_201571.3 NM_201572.3 NM_201593.2 NM_201596.2 NM_201597.2 XM_011519660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5738844 c.11G>A p.C4Y Chr.10: 18261280-18261280 on Build GRCh38
4484382 c.27C>T p.R9R Chr.10: 18261296-18261296 on Build GRCh38
3414886 c.26G>A p.R9H Chr.10: 18261295-18261295 on Build GRCh38
5738843 c.11G>A p.C4Y Chr.10: 18261280-18261280 on Build GRCh38
3741594 c.47A>C p.Y16S Chr.10: 18261316-18261316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997069433 Chr.10: 18261393-18261393 on Build GRCh38
rs149836040 Chr.10: 18261325-18261325 on Build GRCh38
rs977661158 Chr.10: 18261299-18261299 on Build GRCh38
rs1011722727 Chr.10: 18261244-18261244 on Build GRCh38
rs765072163 Chr.10: 18261334-18261334 on Build GRCh38
rs964018178 Chr.10: 18261380-18261380 on Build GRCh38
rs761958542 Chr.10: 18261288-18261288 on Build GRCh38
rs577123395 Chr.10: 18261328-18261328 on Build GRCh38
rs772273592 Chr.10: 18261292-18261292 on Build GRCh38
rs907852635 Chr.10: 18261207-18261207 on Build GRCh38
rs145811885 Chr.10: 18261367-18261367 on Build GRCh38
rs898390924 Chr.10: 18261179-18261179 on Build GRCh38
rs892094980 Chr.10: 18261350-18261350 on Build GRCh38
rs752477754 Chr.10: 18261348-18261348 on Build GRCh38
rs372062152 Chr.10: 18261281-18261281 on Build GRCh38
rs973538244 Chr.10: 18261332-18261332 on Build GRCh38
rs1046997944 Chr.10: 18261205-18261205 on Build GRCh38
rs761633693 Chr.10: 18261368-18261368 on Build GRCh38
rs1002494854 Chr.10: 18261224-18261224 on Build GRCh38
rs374903228 Chr.10: 18261316-18261316 on Build GRCh38
rs749985180 Chr.10: 18261196-18261196 on Build GRCh38
rs786205783 Chr.10: 18261307-18261307 on Build GRCh38
rs967567029 Chr.10: 18261290-18261290 on Build GRCh38
rs112037071 Chr.10: 18261184-18261184 on Build GRCh38
rs1030628843 Chr.10: 18261306-18261306 on Build GRCh38
rs866553442 Chr.10: 18261336-18261336 on Build GRCh38
rs942130254 Chr.10: 18261331-18261331 on Build GRCh38
rs768293777 Chr.10: 18261282-18261282 on Build GRCh38
rs538094186 Chr.10: 18261352-18261352 on Build GRCh38
rs906873969 Chr.10: 18261213-18261213 on Build GRCh38
rs773199951 Chr.10: 18261300-18261300 on Build GRCh38
rs1034067637 Chr.10: 18261225-18261225 on Build GRCh38
rs533673523 Chr.10: 18261302-18261302 on Build GRCh38
rs987568058 Chr.10: 18261194-18261194 on Build GRCh38
rs951045 Chr.10: 18261341-18261341 on Build GRCh38
rs1038325587 Chr.10: 18261217-18261217 on Build GRCh38
rs894227814 Chr.10: 18261242-18261242 on Build GRCh38

View Full Product Details