Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102860107-102860380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS7
Entrez Gene ID
119032
Gene Name
BLOC-1 related complex subunit 7
Gene Aliases
C10orf32
UniGene
Hs.34492
Gene Chromosome Location
Chr.10: 102854210-102864961 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3769065 c.186C>T p.A62A Chr.10: 102860376-102860376 on Build GRCh38
3414643 c.161G>A p.R54Q Chr.10: 102860351-102860351 on Build GRCh38
3433809 c.177G>A p.Q59Q Chr.10: 102860367-102860367 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746676078 Chr.10: 102860229-102860229 on Build GRCh38
rs566553231 Chr.10: 102860206-102860206 on Build GRCh38
rs762780871 Chr.10: 102860350-102860350 on Build GRCh38
rs776210287 Chr.10: 102860302-102860302 on Build GRCh38
rs760396497 Chr.10: 102860287-102860287 on Build GRCh38
rs544397534 Chr.10: 102860245-102860245 on Build GRCh38
rs1001327257 Chr.10: 102860186-102860186 on Build GRCh38
rs34720459 Chr.10: 102860146-102860147 on Build GRCh38
rs533820998 Chr.10: 102860215-102860215 on Build GRCh38
rs932149094 Chr.10: 102860173-102860173 on Build GRCh38
rs1028095203 Chr.10: 102860263-102860263 on Build GRCh38
rs372222571 Chr.10: 102860291-102860291 on Build GRCh38
rs182873252 Chr.10: 102860254-102860254 on Build GRCh38
rs369716429 Chr.10: 102860353-102860353 on Build GRCh38
rs752467994 Chr.10: 102860338-102860338 on Build GRCh38
rs376830356 Chr.10: 102860334-102860334 on Build GRCh38
rs906937848 Chr.10: 102860146-102860146 on Build GRCh38
rs759389843 Chr.10: 102860306-102860306 on Build GRCh38
rs995434748 Chr.10: 102860249-102860249 on Build GRCh38
rs922026695 Chr.10: 102860129-102860129 on Build GRCh38
rs765217405 Chr.10: 102860314-102860314 on Build GRCh38
rs767792654 Chr.10: 102860152-102860152 on Build GRCh38
rs558462398 Chr.10: 102860226-102860226 on Build GRCh38
rs770520865 Chr.10: 102860267-102860267 on Build GRCh38
rs78110962 Chr.10: 102860234-102860234 on Build GRCh38

View Full Product Details