Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12079341-12079602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L14
Entrez Gene ID
79370
Gene Name
BCL2 like 14
Gene Aliases
BCLG
UniGene
Hs.210343
Gene Chromosome Location
Chr.12: 12049844-12099695 on Build GRCh38
Transcripts
NM_030766.1 NM_138722.1 NM_138723.1 XM_006719151.3 XM_011520848.1 XM_011520849.1 XM_017019963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6610710 c.218C>T p.P73L Chr.12: 12079523-12079523 on Build GRCh38
5527310 c.187G>A p.A63T Chr.12: 12079492-12079492 on Build GRCh38
2176250 c.162C>T p.S54S Chr.12: 12079467-12079467 on Build GRCh38
3359588 c.86A>T p.Y29F Chr.12: 12079391-12079391 on Build GRCh38
5374430 c.236C>T p.S79F Chr.12: 12079541-12079541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780055581 Chr.12: 12079575-12079576 on Build GRCh38
rs752451258 Chr.12: 12079480-12079480 on Build GRCh38
rs749596076 Chr.12: 12079546-12079546 on Build GRCh38
rs371514083 Chr.12: 12079472-12079472 on Build GRCh38
rs573345564 Chr.12: 12079575-12079575 on Build GRCh38
rs779436241 Chr.12: 12079566-12079566 on Build GRCh38
rs775821732 Chr.12: 12079434-12079434 on Build GRCh38
rs575710211 Chr.12: 12079458-12079458 on Build GRCh38
rs374164865 Chr.12: 12079476-12079476 on Build GRCh38
rs142096906 Chr.12: 12079387-12079387 on Build GRCh38
rs781260534 Chr.12: 12079402-12079402 on Build GRCh38
rs749449524 Chr.12: 12079442-12079442 on Build GRCh38
rs912194966 Chr.12: 12079433-12079433 on Build GRCh38
rs763879200 Chr.12: 12079496-12079496 on Build GRCh38
rs368530005 Chr.12: 12079487-12079487 on Build GRCh38
rs750292423 Chr.12: 12079570-12079571 on Build GRCh38
rs781069739 Chr.12: 12079513-12079513 on Build GRCh38
rs1043342944 Chr.12: 12079427-12079428 on Build GRCh38
rs759121419 Chr.12: 12079474-12079474 on Build GRCh38
rs61733247 Chr.12: 12079512-12079512 on Build GRCh38
rs748480352 Chr.12: 12079571-12079571 on Build GRCh38
rs769792959 Chr.12: 12079457-12079457 on Build GRCh38
rs141156172 Chr.12: 12079405-12079405 on Build GRCh38
rs758502149 Chr.12: 12079570-12079570 on Build GRCh38
rs376083367 Chr.12: 12079460-12079460 on Build GRCh38
rs745802687 Chr.12: 12079565-12079565 on Build GRCh38
rs140584428 Chr.12: 12079519-12079519 on Build GRCh38
rs750329698 Chr.12: 12079515-12079515 on Build GRCh38
rs142250889 Chr.12: 12079462-12079462 on Build GRCh38
rs769942576 Chr.12: 12079421-12079421 on Build GRCh38
rs756267399 Chr.12: 12079518-12079518 on Build GRCh38
rs61733246 Chr.12: 12079386-12079386 on Build GRCh38
rs61733245 Chr.12: 12079370-12079370 on Build GRCh38
rs772532155 Chr.12: 12079574-12079574 on Build GRCh38
rs1012545639 Chr.12: 12079573-12079573 on Build GRCh38

View Full Product Details