Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101554147-101554380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX1
Entrez Gene ID
51309
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
ALEX1, GASP7
UniGene
Hs.9728
Gene Chromosome Location
Chr.X: 101550531-101554700 on Build GRCh38
Transcripts
NM_016608.1

SNP Detail

SNP ID Chromosome Location
rs781789204 Chr.X: 101554347-101554347 on Build GRCh38
rs782819950 Chr.X: 101554195-101554195 on Build GRCh38
rs200442150 Chr.X: 101554313-101554313 on Build GRCh38
rs147231157 Chr.X: 101554229-101554229 on Build GRCh38
rs1001442610 Chr.X: 101554230-101554230 on Build GRCh38
rs782449508 Chr.X: 101554332-101554332 on Build GRCh38
rs145291885 Chr.X: 101554181-101554181 on Build GRCh38
rs782691299 Chr.X: 101554337-101554337 on Build GRCh38
rs782016786 Chr.X: 101554206-101554206 on Build GRCh38
rs782046340 Chr.X: 101554193-101554193 on Build GRCh38
rs1037559120 Chr.X: 101554223-101554223 on Build GRCh38
rs781935716 Chr.X: 101554190-101554190 on Build GRCh38
rs895819917 Chr.X: 101554338-101554338 on Build GRCh38
rs782638649 Chr.X: 101554297-101554297 on Build GRCh38
rs781821503 Chr.X: 101554331-101554331 on Build GRCh38
rs1055756573 Chr.X: 101554291-101554291 on Build GRCh38
rs782351016 Chr.X: 101554188-101554188 on Build GRCh38

View Full Product Details