Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79977479-79977752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXNL4A
Entrez Gene ID
10907
Gene Name
thioredoxin like 4A
Gene Aliases
BMKS, DIB1, DIM1, SNRNP15, TXNL4, U5-15kD
UniGene
Hs.465498
Gene Chromosome Location
Chr.18: 79972867-80033935 on Build GRCh38
Transcripts
NM_001303471.2 NM_001305557.1 NM_001305563.1 NM_001305564.1 NM_006701.4 NR_131175.1 NR_131176.1 NR_131177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6308376 c.210C>T p.N70N Chr.18: 79977645-79977645 on Build GRCh38
1389872 c.242T>C p.V81A Chr.18: 79977613-79977613 on Build GRCh38
5082879 c.228C>T p.Y76Y Chr.18: 79977627-79977627 on Build GRCh38
6814592 c.226T>C p.Y76H Chr.18: 79977629-79977629 on Build GRCh38
3821770 c.243C>T p.V81V Chr.18: 79977612-79977612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762744869 Chr.18: 79977687-79977687 on Build GRCh38
rs747227156 Chr.18: 79977645-79977645 on Build GRCh38
rs756971133 Chr.18: 79977591-79977591 on Build GRCh38
rs755478919 Chr.18: 79977603-79977603 on Build GRCh38
rs766568151 Chr.18: 79977528-79977528 on Build GRCh38
rs759499936 Chr.18: 79977533-79977533 on Build GRCh38
rs146668986 Chr.18: 79977612-79977612 on Build GRCh38
rs902551697 Chr.18: 79977560-79977560 on Build GRCh38
rs183180579 Chr.18: 79977715-79977715 on Build GRCh38
rs748239471 Chr.18: 79977575-79977576 on Build GRCh38
rs776825515 Chr.18: 79977663-79977663 on Build GRCh38
rs771287209 Chr.18: 79977715-79977716 on Build GRCh38
rs760777183 Chr.18: 79977525-79977525 on Build GRCh38
rs758153246 Chr.18: 79977561-79977561 on Build GRCh38
rs773499997 Chr.18: 79977639-79977639 on Build GRCh38
rs934716212 Chr.18: 79977695-79977695 on Build GRCh38
rs201489532 Chr.18: 79977627-79977627 on Build GRCh38
rs765175716 Chr.18: 79977675-79977675 on Build GRCh38
rs112183663 Chr.18: 79977705-79977705 on Build GRCh38
rs759569966 Chr.18: 79977671-79977671 on Build GRCh38
rs563943079 Chr.18: 79977588-79977588 on Build GRCh38
rs545058138 Chr.18: 79977559-79977559 on Build GRCh38
rs777427941 Chr.18: 79977585-79977585 on Build GRCh38
rs763874928 Chr.18: 79977696-79977696 on Build GRCh38
rs754034313 Chr.18: 79977529-79977529 on Build GRCh38
rs148835299 Chr.18: 79977621-79977621 on Build GRCh38
rs772315346 Chr.18: 79977715-79977715 on Build GRCh38
rs773453340 Chr.18: 79977714-79977715 on Build GRCh38
rs771333931 Chr.18: 79977654-79977654 on Build GRCh38
rs778777844 Chr.18: 79977525-79977525 on Build GRCh38
rs772296279 Chr.18: 79977521-79977521 on Build GRCh38
rs773588334 Chr.18: 79977523-79977523 on Build GRCh38
rs530980053 Chr.18: 79977597-79977597 on Build GRCh38
rs780667220 Chr.18: 79977593-79977593 on Build GRCh38
rs762124416 Chr.18: 79977511-79977511 on Build GRCh38
rs368956183 Chr.18: 79977556-79977556 on Build GRCh38
rs775306959 Chr.18: 79977678-79977678 on Build GRCh38

View Full Product Details