Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 84603765-84604034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN2
Entrez Gene ID
54993
Gene Name
zinc finger and SCAN domain containing 2
Gene Aliases
ZFP29, ZNF854
UniGene
Hs.594023
Gene Chromosome Location
Chr.15: 84600986-84623716 on Build GRCh38
Transcripts
NM_001007072.1 NM_017894.5 NM_181877.3 XM_017022392.1 XM_017022393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6834285 c.21G>A p.P7P Chr.15: 84603948-84603948 on Build GRCh38
5091405 c.36G>A p.P12P Chr.15: 84603963-84603963 on Build GRCh38
1375054 c.36G>A p.P12P Chr.15: 84603963-84603963 on Build GRCh38
3504717 c.102C>T p.I34I Chr.15: 84604029-84604029 on Build GRCh38
3504714 c.79G>A p.E27K Chr.15: 84604006-84604006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760416487 Chr.15: 84603996-84603996 on Build GRCh38
rs747332308 Chr.15: 84603959-84603959 on Build GRCh38
rs367963102 Chr.15: 84603963-84603963 on Build GRCh38
rs771595597 Chr.15: 84603961-84603961 on Build GRCh38
rs916101873 Chr.15: 84603990-84603990 on Build GRCh38
rs777228309 Chr.15: 84603962-84603962 on Build GRCh38
rs745946681 Chr.15: 84604004-84604006 on Build GRCh38
rs150032800 Chr.15: 84603799-84603799 on Build GRCh38
rs773223840 Chr.15: 84604000-84604003 on Build GRCh38
rs779928801 Chr.15: 84603941-84603941 on Build GRCh38
rs771738448 Chr.15: 84603949-84603949 on Build GRCh38
rs778011907 Chr.15: 84603958-84603958 on Build GRCh38
rs770068661 Chr.15: 84603964-84603964 on Build GRCh38
rs112520506 Chr.15: 84603857-84603857 on Build GRCh38
rs573463675 Chr.15: 84603867-84603867 on Build GRCh38
rs146866915 Chr.15: 84603947-84603947 on Build GRCh38
rs997550737 Chr.15: 84603883-84603883 on Build GRCh38
rs965096812 Chr.15: 84603796-84603796 on Build GRCh38
rs772915822 Chr.15: 84603995-84603995 on Build GRCh38
rs375266737 Chr.15: 84603977-84603977 on Build GRCh38
rs555519185 Chr.15: 84603903-84603903 on Build GRCh38
rs535802249 Chr.15: 84603887-84603887 on Build GRCh38
rs544746857 Chr.15: 84603979-84603979 on Build GRCh38
rs747817109 Chr.15: 84603948-84603948 on Build GRCh38
rs924725525 Chr.15: 84603952-84603952 on Build GRCh38
rs763097992 Chr.15: 84603971-84603971 on Build GRCh38
rs915111589 Chr.15: 84603914-84603914 on Build GRCh38
rs530227193 Chr.15: 84603861-84603861 on Build GRCh38
rs140727399 Chr.15: 84604004-84604004 on Build GRCh38
rs202060203 Chr.15: 84603999-84603999 on Build GRCh38
rs754901664 Chr.15: 84604015-84604015 on Build GRCh38
rs372556238 Chr.15: 84603966-84603966 on Build GRCh38
rs375993423 Chr.15: 84603856-84603856 on Build GRCh38
rs753809090 Chr.15: 84604014-84604014 on Build GRCh38
rs754528042 Chr.15: 84603945-84603945 on Build GRCh38
rs1026383976 Chr.15: 84603901-84603901 on Build GRCh38
rs147730851 Chr.15: 84603821-84603821 on Build GRCh38
rs901054728 Chr.15: 84603854-84603854 on Build GRCh38
rs968751907 Chr.15: 84603915-84603915 on Build GRCh38
rs182736722 Chr.15: 84603944-84603944 on Build GRCh38
rs750177411 Chr.15: 84603923-84603923 on Build GRCh38
rs989428844 Chr.15: 84603864-84603864 on Build GRCh38
rs756013915 Chr.15: 84603930-84603930 on Build GRCh38
rs1040819848 Chr.15: 84603819-84603819 on Build GRCh38

View Full Product Details