Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169596600-169596839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDL1
Entrez Gene ID
54908
Gene Name
spindle apparatus coiled-coil protein 1
Gene Aliases
CCDC99
UniGene
Hs.368710
Gene Chromosome Location
Chr.5: 169583634-169604778 on Build GRCh38
Transcripts
NM_017785.4 XM_011534585.1 XM_011534586.2 XM_011534587.2 XM_011534588.2 XM_011534589.2 XM_011534590.2 XM_017009601.1 XM_017009602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
736812 c.948G>T p.Q316H Chr.5: 169596617-169596617 on Build GRCh38
5505889 c.1025A>C p.K342T Chr.5: 169596694-169596694 on Build GRCh38
3338821 c.1021G>A p.E341K Chr.5: 169596690-169596690 on Build GRCh38
6102963 c.936A>G p.E312E Chr.5: 169596605-169596605 on Build GRCh38
1065684 c.1018C>A p.L340M Chr.5: 169596687-169596687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779833644 Chr.5: 169596720-169596720 on Build GRCh38
rs767438458 Chr.5: 169596729-169596729 on Build GRCh38
rs568119516 Chr.5: 169596723-169596724 on Build GRCh38
rs774516530 Chr.5: 169596666-169596666 on Build GRCh38
rs575676623 Chr.5: 169596713-169596713 on Build GRCh38
rs756263190 Chr.5: 169596717-169596717 on Build GRCh38
rs779431833 Chr.5: 169596732-169596733 on Build GRCh38
rs759835725 Chr.5: 169596668-169596668 on Build GRCh38
rs182736283 Chr.5: 169596734-169596734 on Build GRCh38
rs767861778 Chr.5: 169596678-169596678 on Build GRCh38
rs200981583 Chr.5: 169596699-169596699 on Build GRCh38
rs753379535 Chr.5: 169596698-169596698 on Build GRCh38
rs772534639 Chr.5: 169596740-169596740 on Build GRCh38
rs771109869 Chr.5: 169596662-169596662 on Build GRCh38
rs749401424 Chr.5: 169596741-169596741 on Build GRCh38
rs765016800 Chr.5: 169596702-169596702 on Build GRCh38
rs1055091297 Chr.5: 169596785-169596785 on Build GRCh38
rs570682508 Chr.5: 169596717-169596719 on Build GRCh38
rs763757135 Chr.5: 169596687-169596687 on Build GRCh38
rs936389496 Chr.5: 169596705-169596705 on Build GRCh38
rs144297297 Chr.5: 169596682-169596682 on Build GRCh38
rs764216296 Chr.5: 169596776-169596776 on Build GRCh38
rs147810277 Chr.5: 169596684-169596684 on Build GRCh38
rs142021941 Chr.5: 169596621-169596621 on Build GRCh38
rs370549413 Chr.5: 169596724-169596724 on Build GRCh38
rs1002031032 Chr.5: 169596772-169596772 on Build GRCh38
rs754144541 Chr.5: 169596716-169596717 on Build GRCh38
rs906372660 Chr.5: 169596747-169596747 on Build GRCh38
rs748592413 Chr.5: 169596736-169596736 on Build GRCh38
rs758143100 Chr.5: 169596717-169596717 on Build GRCh38
rs773264345 Chr.5: 169596633-169596633 on Build GRCh38
rs190653043 Chr.5: 169596680-169596680 on Build GRCh38
rs749559116 Chr.5: 169596647-169596647 on Build GRCh38

View Full Product Details