Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6856168-6856441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP5
Entrez Gene ID
8078
Gene Name
ubiquitin specific peptidase 5
Gene Aliases
ISOT
UniGene
Hs.631661
Gene Chromosome Location
Chr.12: 6852118-6866632 on Build GRCh38
Transcripts
NM_001098536.1 NM_003481.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1363952 c.572G>A p.R191Q Chr.12: 6856438-6856438 on Build GRCh38
7329124 c.439G>T p.V147L Chr.12: 6856305-6856305 on Build GRCh38
1363953 c.572G>A p.R191Q Chr.12: 6856438-6856438 on Build GRCh38
1676681 c.520C>T p.R174W Chr.12: 6856386-6856386 on Build GRCh38
1676680 c.520C>T p.R174W Chr.12: 6856386-6856386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781916966 Chr.12: 6856301-6856301 on Build GRCh38
rs781878572 Chr.12: 6856353-6856353 on Build GRCh38
rs752595305 Chr.12: 6856350-6856350 on Build GRCh38
rs185830650 Chr.12: 6856338-6856338 on Build GRCh38
rs575404753 Chr.12: 6856418-6856418 on Build GRCh38
rs782298280 Chr.12: 6856262-6856262 on Build GRCh38
rs781898760 Chr.12: 6856233-6856233 on Build GRCh38
rs782293734 Chr.12: 6856393-6856393 on Build GRCh38
rs373122898 Chr.12: 6856334-6856334 on Build GRCh38
rs199642135 Chr.12: 6856273-6856273 on Build GRCh38
rs782076351 Chr.12: 6856403-6856403 on Build GRCh38
rs558740385 Chr.12: 6856379-6856379 on Build GRCh38
rs782210685 Chr.12: 6856290-6856290 on Build GRCh38
rs782031903 Chr.12: 6856319-6856319 on Build GRCh38
rs782324656 Chr.12: 6856387-6856387 on Build GRCh38
rs190200818 Chr.12: 6856391-6856391 on Build GRCh38
rs782606734 Chr.12: 6856251-6856251 on Build GRCh38
rs782672954 Chr.12: 6856254-6856254 on Build GRCh38
rs781893442 Chr.12: 6856337-6856337 on Build GRCh38
rs115270213 Chr.12: 6856244-6856244 on Build GRCh38
rs781816036 Chr.12: 6856330-6856330 on Build GRCh38
rs782095332 Chr.12: 6856333-6856333 on Build GRCh38
rs782021481 Chr.12: 6856320-6856320 on Build GRCh38
rs782627007 Chr.12: 6856383-6856383 on Build GRCh38
rs782772619 Chr.12: 6856326-6856326 on Build GRCh38
rs140761461 Chr.12: 6856411-6856411 on Build GRCh38
rs782791191 Chr.12: 6856312-6856312 on Build GRCh38
rs782639190 Chr.12: 6856352-6856352 on Build GRCh38
rs782471816 Chr.12: 6856382-6856382 on Build GRCh38
rs200472768 Chr.12: 6856386-6856386 on Build GRCh38
rs782578478 Chr.12: 6856307-6856307 on Build GRCh38
rs782182525 Chr.12: 6856198-6856198 on Build GRCh38
rs781998201 Chr.12: 6856400-6856400 on Build GRCh38
rs782370913 Chr.12: 6856395-6856395 on Build GRCh38
rs782458438 Chr.12: 6856199-6856199 on Build GRCh38
rs143386381 Chr.12: 6856268-6856268 on Build GRCh38
rs201361484 Chr.12: 6856295-6856295 on Build GRCh38
rs1049486177 Chr.12: 6856420-6856420 on Build GRCh38

View Full Product Details