Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39120990-39121261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC21A
Entrez Gene ID
199223
Gene Name
tetratricopeptide repeat domain 21A
Gene Aliases
IFT139A, STI2
UniGene
Hs.443935
Gene Chromosome Location
Chr.3: 39107661-39139009 on Build GRCh38
Transcripts
NM_001105513.2 NM_145755.2 XM_005264921.4 XM_005264922.4 XM_005264923.3 XM_005264924.4 XM_005264925.4 XM_005264926.4 XM_005264927.4 XM_006713011.3 XM_011533447.2 XM_011533449.1 XM_017005843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6097745 c.1103_1104TG>AT p.M368N Chr.3: 39121175-39121176 on Build GRCh38
1044425 c.1063C>A p.L355M Chr.3: 39121135-39121135 on Build GRCh38
6544359 c.990C>T p.T330T Chr.3: 39121062-39121062 on Build GRCh38
7181494 c.1029C>T p.L343L Chr.3: 39121101-39121101 on Build GRCh38
4896925 c.1015G>A p.E339K Chr.3: 39121087-39121087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767289348 Chr.3: 39121160-39121160 on Build GRCh38
rs368270092 Chr.3: 39121227-39121227 on Build GRCh38
rs369704736 Chr.3: 39121041-39121041 on Build GRCh38
rs374496008 Chr.3: 39121187-39121187 on Build GRCh38
rs758671378 Chr.3: 39121182-39121182 on Build GRCh38
rs202013731 Chr.3: 39121106-39121106 on Build GRCh38
rs748687254 Chr.3: 39121195-39121195 on Build GRCh38
rs777202225 Chr.3: 39121105-39121105 on Build GRCh38
rs764738378 Chr.3: 39121027-39121027 on Build GRCh38
rs754883644 Chr.3: 39121068-39121068 on Build GRCh38
rs777094369 Chr.3: 39121140-39121140 on Build GRCh38
rs756062919 Chr.3: 39121162-39121162 on Build GRCh38
rs762565595 Chr.3: 39121131-39121131 on Build GRCh38
rs367889160 Chr.3: 39121077-39121077 on Build GRCh38
rs776053682 Chr.3: 39121011-39121011 on Build GRCh38
rs770247738 Chr.3: 39121199-39121199 on Build GRCh38
rs772489402 Chr.3: 39121082-39121082 on Build GRCh38
rs747400934 Chr.3: 39121093-39121093 on Build GRCh38
rs571677940 Chr.3: 39121242-39121242 on Build GRCh38
rs766072904 Chr.3: 39121133-39121133 on Build GRCh38
rs374133009 Chr.3: 39121155-39121155 on Build GRCh38
rs563644994 Chr.3: 39121009-39121009 on Build GRCh38
rs796655541 Chr.3: 39121067-39121067 on Build GRCh38
rs753818179 Chr.3: 39121071-39121071 on Build GRCh38
rs201536754 Chr.3: 39121070-39121070 on Build GRCh38
rs780534542 Chr.3: 39121083-39121083 on Build GRCh38
rs746088486 Chr.3: 39121081-39121081 on Build GRCh38
rs750107530 Chr.3: 39121045-39121045 on Build GRCh38
rs775956871 Chr.3: 39121040-39121040 on Build GRCh38
rs944520204 Chr.3: 39121099-39121099 on Build GRCh38
rs368492109 Chr.3: 39121239-39121239 on Build GRCh38
rs751303690 Chr.3: 39121139-39121139 on Build GRCh38
rs772666910 Chr.3: 39121129-39121129 on Build GRCh38
rs921796101 Chr.3: 39121137-39121137 on Build GRCh38
rs781470353 Chr.3: 39121191-39121191 on Build GRCh38
rs780455168 Chr.3: 39121183-39121183 on Build GRCh38
rs769156838 Chr.3: 39121190-39121190 on Build GRCh38
rs757360106 Chr.3: 39121075-39121075 on Build GRCh38
rs760087289 Chr.3: 39121173-39121173 on Build GRCh38
rs751399928 Chr.3: 39121050-39121050 on Build GRCh38
rs769337456 Chr.3: 39121128-39121128 on Build GRCh38
rs752448296 Chr.3: 39121164-39121164 on Build GRCh38
rs765217315 Chr.3: 39121127-39121128 on Build GRCh38
rs868243044 Chr.3: 39121104-39121104 on Build GRCh38
rs769246568 Chr.3: 39121102-39121102 on Build GRCh38
rs764731030 Chr.3: 39121042-39121042 on Build GRCh38
rs199791048 Chr.3: 39121178-39121178 on Build GRCh38
rs373055793 Chr.3: 39121069-39121069 on Build GRCh38
rs758047012 Chr.3: 39121046-39121046 on Build GRCh38
rs199538696 Chr.3: 39121161-39121161 on Build GRCh38
rs551865452 Chr.3: 39121200-39121200 on Build GRCh38
rs926216536 Chr.3: 39121044-39121044 on Build GRCh38
rs372844330 Chr.3: 39121226-39121226 on Build GRCh38
rs368573540 Chr.3: 39121202-39121202 on Build GRCh38
rs766162671 Chr.3: 39121048-39121048 on Build GRCh38
rs773145535 Chr.3: 39121191-39121192 on Build GRCh38
rs533681271 Chr.3: 39121238-39121238 on Build GRCh38
rs761260045 Chr.3: 39121012-39121012 on Build GRCh38
rs767249879 Chr.3: 39121087-39121087 on Build GRCh38

View Full Product Details