Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42581309-42581558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC30
Entrez Gene ID
728621
Gene Name
coiled-coil domain containing 30
Gene Aliases
PFD6L, PFDN6L
UniGene
Hs.729640
Gene Chromosome Location
Chr.1: 42521240-42654664 on Build GRCh38
Transcripts
NM_001080850.2 XM_011542067.2 XM_011542068.2 XM_011542069.2 XM_011542070.2 XM_011542071.2 XM_017002225.1 XM_017002226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4238218 c.485A>C p.D162A Chr.1: 42581412-42581412 on Build GRCh38
4419052 c.1092A>G p.E364E Chr.1: 42581386-42581386 on Build GRCh38
6398971 c.1181C>A p.A394D Chr.1: 42581475-42581475 on Build GRCh38
5732495 c.1072G>T p.D358Y Chr.1: 42581366-42581366 on Build GRCh38
6398972 c.548C>A p.A183D Chr.1: 42581475-42581475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748517760 Chr.1: 42581395-42581395 on Build GRCh38
rs139002601 Chr.1: 42581438-42581438 on Build GRCh38
rs1034231516 Chr.1: 42581375-42581375 on Build GRCh38
rs753635743 Chr.1: 42581482-42581482 on Build GRCh38
rs765148066 Chr.1: 42581493-42581493 on Build GRCh38
rs763421494 Chr.1: 42581416-42581416 on Build GRCh38
rs369659878 Chr.1: 42581524-42581524 on Build GRCh38
rs767853148 Chr.1: 42581455-42581455 on Build GRCh38
rs779625472 Chr.1: 42581393-42581393 on Build GRCh38
rs770018297 Chr.1: 42581396-42581396 on Build GRCh38
rs760326933 Chr.1: 42581349-42581349 on Build GRCh38
rs773795387 Chr.1: 42581407-42581407 on Build GRCh38
rs917309716 Chr.1: 42581391-42581391 on Build GRCh38
rs771440007 Chr.1: 42581418-42581418 on Build GRCh38
rs753668559 Chr.1: 42581368-42581368 on Build GRCh38
rs920576172 Chr.1: 42581490-42581490 on Build GRCh38
rs12037518 Chr.1: 42581386-42581386 on Build GRCh38
rs202069590 Chr.1: 42581373-42581373 on Build GRCh38
rs1021940349 Chr.1: 42581356-42581356 on Build GRCh38
rs771771878 Chr.1: 42581388-42581388 on Build GRCh38
rs760539985 Chr.1: 42581462-42581462 on Build GRCh38
rs868214289 Chr.1: 42581471-42581471 on Build GRCh38
rs757837692 Chr.1: 42581503-42581503 on Build GRCh38
rs140773357 Chr.1: 42581376-42581376 on Build GRCh38
rs749950787 Chr.1: 42581497-42581497 on Build GRCh38
rs757207322 Chr.1: 42581489-42581489 on Build GRCh38
rs777793972 Chr.1: 42581360-42581360 on Build GRCh38
rs752550386 Chr.1: 42581341-42581341 on Build GRCh38
rs763843716 Chr.1: 42581472-42581472 on Build GRCh38
rs755794589 Chr.1: 42581346-42581346 on Build GRCh38
rs759628119 Chr.1: 42581449-42581449 on Build GRCh38
rs996230610 Chr.1: 42581352-42581352 on Build GRCh38
rs375870892 Chr.1: 42581409-42581409 on Build GRCh38
rs1030927819 Chr.1: 42581374-42581374 on Build GRCh38
rs775810460 Chr.1: 42581457-42581457 on Build GRCh38

View Full Product Details