Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63477978-63478233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACE
Entrez Gene ID
1636
Gene Name
angiotensin I converting enzyme
Gene Aliases
ACE1, CD143, DCP, DCP1, ICH, MVCD3
UniGene
Hs.298469
Gene Chromosome Location
Chr.17: 63477061-63498380 on Build GRCh38
Transcripts
NM_000789.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6386081 c.386C>A p.S129Y Chr.17: 63478067-63478067 on Build GRCh38
6386082 c.386C>A p.S129Y Chr.17: 63478067-63478067 on Build GRCh38
5387005 c.300G>A p.K100K Chr.17: 63477981-63477981 on Build GRCh38
5962566 c.339G>C p.T113T Chr.17: 63478020-63478020 on Build GRCh38
6081824 c.312G>T p.L104L Chr.17: 63477993-63477993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779422412 Chr.17: 63478093-63478093 on Build GRCh38
rs770863321 Chr.17: 63478040-63478040 on Build GRCh38
rs764488884 Chr.17: 63478090-63478090 on Build GRCh38
rs770787358 Chr.17: 63478117-63478117 on Build GRCh38
rs904130482 Chr.17: 63478055-63478055 on Build GRCh38
rs760357465 Chr.17: 63478140-63478140 on Build GRCh38
rs1015502824 Chr.17: 63478125-63478125 on Build GRCh38
rs772201818 Chr.17: 63478001-63478001 on Build GRCh38
rs541197581 Chr.17: 63478115-63478115 on Build GRCh38
rs114000559 Chr.17: 63478106-63478106 on Build GRCh38
rs1024071710 Chr.17: 63478105-63478105 on Build GRCh38
rs759391801 Chr.17: 63478129-63478129 on Build GRCh38
rs774282606 Chr.17: 63478121-63478121 on Build GRCh38
rs768970823 Chr.17: 63478110-63478110 on Build GRCh38
rs758113296 Chr.17: 63478182-63478182 on Build GRCh38
rs780364983 Chr.17: 63478011-63478011 on Build GRCh38
rs767085054 Chr.17: 63478064-63478064 on Build GRCh38
rs750481872 Chr.17: 63478096-63478096 on Build GRCh38
rs1028196239 Chr.17: 63478207-63478207 on Build GRCh38
rs747960753 Chr.17: 63478027-63478027 on Build GRCh38
rs754220619 Chr.17: 63478156-63478156 on Build GRCh38
rs936814960 Chr.17: 63478058-63478058 on Build GRCh38
rs556087296 Chr.17: 63478091-63478091 on Build GRCh38
rs374143795 Chr.17: 63478141-63478141 on Build GRCh38
rs369526392 Chr.17: 63478142-63478142 on Build GRCh38
rs539067889 Chr.17: 63478042-63478042 on Build GRCh38
rs145692547 Chr.17: 63478030-63478030 on Build GRCh38
rs3729659 Chr.17: 63478092-63478092 on Build GRCh38
rs564425051 Chr.17: 63478130-63478130 on Build GRCh38
rs773121538 Chr.17: 63478034-63478034 on Build GRCh38
rs945596173 Chr.17: 63478208-63478208 on Build GRCh38
rs561973858 Chr.17: 63478108-63478108 on Build GRCh38
rs3729660 Chr.17: 63478111-63478111 on Build GRCh38
rs774962048 Chr.17: 63478131-63478131 on Build GRCh38
rs749921989 Chr.17: 63478180-63478180 on Build GRCh38
rs762839851 Chr.17: 63478039-63478039 on Build GRCh38
rs759033270 Chr.17: 63478054-63478054 on Build GRCh38
rs752416873 Chr.17: 63478084-63478084 on Build GRCh38
rs758436458 Chr.17: 63478102-63478102 on Build GRCh38
rs1055333381 Chr.17: 63478065-63478065 on Build GRCh38
rs760310248 Chr.17: 63478088-63478088 on Build GRCh38

View Full Product Details