Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21304871-21305138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNA5
Entrez Gene ID
3442
Gene Name
interferon alpha 5
Gene Aliases
IFN-alpha-5, IFN-alphaG, INA5, INFA5, leIF G
UniGene
Hs.37113
Gene Chromosome Location
Chr.9: 21304614-21305313 on Build GRCh38
Transcripts
NM_002169.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291576 c.350_351insA p.N117fs*2 Chr.9: 21304906-21304907 on Build GRCh38
311922 c.254A>T p.Q85L Chr.9: 21305003-21305003 on Build GRCh38
6220636 c.328A>G p.T110A Chr.9: 21304929-21304929 on Build GRCh38
6247293 c.184C>A p.P62T Chr.9: 21305073-21305073 on Build GRCh38
3656580 c.240C>T p.L80L Chr.9: 21305017-21305017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765147470 Chr.9: 21304967-21304967 on Build GRCh38
rs532398110 Chr.9: 21305041-21305041 on Build GRCh38
rs758221007 Chr.9: 21304895-21304895 on Build GRCh38
rs747491497 Chr.9: 21305017-21305017 on Build GRCh38
rs750110646 Chr.9: 21305001-21305001 on Build GRCh38
rs138121530 Chr.9: 21305049-21305049 on Build GRCh38
rs974112792 Chr.9: 21304949-21304949 on Build GRCh38
rs547216857 Chr.9: 21305073-21305073 on Build GRCh38
rs761772402 Chr.9: 21305043-21305043 on Build GRCh38
rs766337836 Chr.9: 21305034-21305034 on Build GRCh38
rs777394094 Chr.9: 21304896-21304896 on Build GRCh38
rs777005860 Chr.9: 21304957-21304957 on Build GRCh38
rs199681254 Chr.9: 21304905-21304905 on Build GRCh38
rs780616072 Chr.9: 21305000-21305000 on Build GRCh38
rs778128018 Chr.9: 21305010-21305010 on Build GRCh38
rs757817708 Chr.9: 21305071-21305071 on Build GRCh38
rs768371402 Chr.9: 21305086-21305086 on Build GRCh38
rs776811060 Chr.9: 21305025-21305025 on Build GRCh38
rs568144733 Chr.9: 21305101-21305101 on Build GRCh38
rs748709351 Chr.9: 21304923-21304923 on Build GRCh38
rs759888184 Chr.9: 21305034-21305034 on Build GRCh38
rs41298198 Chr.9: 21304914-21304914 on Build GRCh38
rs752685144 Chr.9: 21304893-21304893 on Build GRCh38
rs201257845 Chr.9: 21305076-21305076 on Build GRCh38
rs751003739 Chr.9: 21304899-21304899 on Build GRCh38
rs780812383 Chr.9: 21304903-21304903 on Build GRCh38
rs779296896 Chr.9: 21305004-21305004 on Build GRCh38
rs149567637 Chr.9: 21305060-21305060 on Build GRCh38
rs950531422 Chr.9: 21304962-21304962 on Build GRCh38
rs563423510 Chr.9: 21305039-21305039 on Build GRCh38
rs140371188 Chr.9: 21304892-21304892 on Build GRCh38
rs759764056 Chr.9: 21304961-21304961 on Build GRCh38
rs771137726 Chr.9: 21305021-21305021 on Build GRCh38
rs184880532 Chr.9: 21305072-21305072 on Build GRCh38
rs775591446 Chr.9: 21305081-21305081 on Build GRCh38
rs148682805 Chr.9: 21304979-21304979 on Build GRCh38
rs541092287 Chr.9: 21304901-21304901 on Build GRCh38
rs762988389 Chr.9: 21304982-21304982 on Build GRCh38
rs543743851 Chr.9: 21305002-21305002 on Build GRCh38
rs140936266 Chr.9: 21305094-21305094 on Build GRCh38
rs755801741 Chr.9: 21305051-21305051 on Build GRCh38
rs748426399 Chr.9: 21305005-21305005 on Build GRCh38
rs752519749 Chr.9: 21304972-21304972 on Build GRCh38
rs760161253 Chr.9: 21305104-21305104 on Build GRCh38
rs373247063 Chr.9: 21304911-21304911 on Build GRCh38
rs559095007 Chr.9: 21305058-21305058 on Build GRCh38
rs749463200 Chr.9: 21305085-21305085 on Build GRCh38
rs753160327 Chr.9: 21305055-21305055 on Build GRCh38
rs774150373 Chr.9: 21305088-21305088 on Build GRCh38
rs371906885 Chr.9: 21305054-21305054 on Build GRCh38
rs769493970 Chr.9: 21305008-21305008 on Build GRCh38
rs756735402 Chr.9: 21304999-21304999 on Build GRCh38
rs763935604 Chr.9: 21304989-21304989 on Build GRCh38
rs773376794 Chr.9: 21304944-21304944 on Build GRCh38
rs368215360 Chr.9: 21305006-21305006 on Build GRCh38
rs200585197 Chr.9: 21304951-21304951 on Build GRCh38
rs761784153 Chr.9: 21305092-21305092 on Build GRCh38
rs771241699 Chr.9: 21304953-21304953 on Build GRCh38
rs772434166 Chr.9: 21304929-21304929 on Build GRCh38
rs775423049 Chr.9: 21305037-21305037 on Build GRCh38
rs760663380 Chr.9: 21304945-21304945 on Build GRCh38
rs747500740 Chr.9: 21305069-21305069 on Build GRCh38
rs371183306 Chr.9: 21305074-21305074 on Build GRCh38
rs751142857 Chr.9: 21304997-21304997 on Build GRCh38
rs770111825 Chr.9: 21305036-21305036 on Build GRCh38

View Full Product Details