Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43407545-43407782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
UniGene
Hs.164453
Gene Chromosome Location
Chr.7: 43112580-43566001 on Build GRCh38
Transcripts
NM_001287059.1 NM_015052.4 XM_005249665.3 XM_006715670.3 XM_006715671.3 XM_006715673.3 XM_011515220.2 XM_011515222.2 XM_011515223.2 XM_011515224.2 XM_011515225.2 XM_011515226.2 XM_017011882.1 XM_017011883.1 XM_017011884.1 XM_017011885.1 XM_017011886.1 XM_017011887.1 XM_017011888.1 XM_017011889.1 XM_017011890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926594 c.728C>T p.A243V Chr.7: 43407658-43407658 on Build GRCh38
1254070 c.653A>C p.K218T Chr.7: 43407583-43407583 on Build GRCh38
1450779 c.797C>T p.A266V Chr.7: 43407727-43407727 on Build GRCh38
3638748 c.730C>T p.L244F Chr.7: 43407660-43407660 on Build GRCh38
1450777 c.664T>C p.F222L Chr.7: 43407594-43407594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021180141 Chr.7: 43407697-43407697 on Build GRCh38
rs751791426 Chr.7: 43407749-43407749 on Build GRCh38
rs768209619 Chr.7: 43407676-43407676 on Build GRCh38
rs200458933 Chr.7: 43407656-43407656 on Build GRCh38
rs371180560 Chr.7: 43407703-43407703 on Build GRCh38
rs1019383705 Chr.7: 43407671-43407671 on Build GRCh38
rs767043806 Chr.7: 43407689-43407689 on Build GRCh38
rs201174011 Chr.7: 43407584-43407584 on Build GRCh38
rs749035053 Chr.7: 43407668-43407668 on Build GRCh38
rs199623088 Chr.7: 43407741-43407741 on Build GRCh38
rs201297726 Chr.7: 43407708-43407708 on Build GRCh38
rs760092845 Chr.7: 43407707-43407707 on Build GRCh38
rs997693607 Chr.7: 43407688-43407688 on Build GRCh38
rs541016626 Chr.7: 43407608-43407608 on Build GRCh38
rs757352094 Chr.7: 43407614-43407614 on Build GRCh38
rs202123661 Chr.7: 43407638-43407638 on Build GRCh38
rs745781767 Chr.7: 43407657-43407657 on Build GRCh38
rs757329132 Chr.7: 43407751-43407751 on Build GRCh38
rs773989646 Chr.7: 43407679-43407679 on Build GRCh38
rs756505360 Chr.7: 43407750-43407751 on Build GRCh38
rs753122454 Chr.7: 43407728-43407728 on Build GRCh38
rs559436029 Chr.7: 43407680-43407680 on Build GRCh38
rs758765722 Chr.7: 43407731-43407731 on Build GRCh38
rs866714710 Chr.7: 43407581-43407581 on Build GRCh38
rs930939044 Chr.7: 43407655-43407655 on Build GRCh38
rs775295849 Chr.7: 43407663-43407663 on Build GRCh38
rs765786632 Chr.7: 43407718-43407718 on Build GRCh38

View Full Product Details