Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136805951-136806224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC183
Entrez Gene ID
84960
Gene Name
coiled-coil domain containing 183
Gene Aliases
KIAA1984, PARF, bA216L13.7
UniGene
Hs.728796
Gene Chromosome Location
Chr.9: 136796338-136807741 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1

Gene Symbol
CCDC183-AS1
Entrez Gene ID
100131193
Gene Name
CCDC183 antisense RNA 1
Gene Aliases
KIAA1984-AS1
UniGene
Hs.734812
Gene Chromosome Location
Chr.9: 136803927-136808848 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4442273 c.990G>T p.E330D Chr.9: 136806119-136806119 on Build GRCh38
4816680 c.1068G>A p.E356E Chr.9: 136806197-136806197 on Build GRCh38
5646498 c.1080C>G p.L360L Chr.9: 136806209-136806209 on Build GRCh38
4163579 c.1024T>C p.W342R Chr.9: 136806153-136806153 on Build GRCh38
4163580 c.1031T>G p.V344G Chr.9: 136806160-136806160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781190127 Chr.9: 136806156-136806157 on Build GRCh38
rs968069452 Chr.9: 136806107-136806107 on Build GRCh38
rs780118428 Chr.9: 136806164-136806164 on Build GRCh38
rs553757158 Chr.9: 136806113-136806113 on Build GRCh38
rs868370991 Chr.9: 136806106-136806106 on Build GRCh38
rs948076206 Chr.9: 136806061-136806061 on Build GRCh38
rs1029437883 Chr.9: 136806090-136806090 on Build GRCh38
rs370023138 Chr.9: 136805978-136805978 on Build GRCh38
rs765834795 Chr.9: 136806157-136806157 on Build GRCh38
rs772118361 Chr.9: 136806067-136806067 on Build GRCh38
rs770286625 Chr.9: 136806139-136806168 on Build GRCh38
rs748087708 Chr.9: 136806048-136806048 on Build GRCh38
rs753485911 Chr.9: 136806159-136806159 on Build GRCh38
rs778850454 Chr.9: 136806161-136806161 on Build GRCh38
rs185673307 Chr.9: 136806072-136806072 on Build GRCh38
rs775651569 Chr.9: 136806089-136806089 on Build GRCh38
rs1052477300 Chr.9: 136806112-136806112 on Build GRCh38
rs901956114 Chr.9: 136806064-136806064 on Build GRCh38
rs763168465 Chr.9: 136806091-136806091 on Build GRCh38
rs920540355 Chr.9: 136806013-136806013 on Build GRCh38
rs2811795 Chr.9: 136806153-136806153 on Build GRCh38
rs769786981 Chr.9: 136806083-136806083 on Build GRCh38
rs749497442 Chr.9: 136806177-136806177 on Build GRCh38
rs762222241 Chr.9: 136806141-136806141 on Build GRCh38
rs201907900 Chr.9: 136806204-136806204 on Build GRCh38
rs759943438 Chr.9: 136806027-136806027 on Build GRCh38
rs1005017668 Chr.9: 136806003-136806003 on Build GRCh38
rs749875462 Chr.9: 136806154-136806154 on Build GRCh38
rs201957004 Chr.9: 136806186-136806186 on Build GRCh38
rs574486749 Chr.9: 136806066-136806066 on Build GRCh38
rs973309086 Chr.9: 136805984-136805984 on Build GRCh38
rs769847585 Chr.9: 136806163-136806163 on Build GRCh38
rs758247195 Chr.9: 136806162-136806162 on Build GRCh38
rs764382554 Chr.9: 136806110-136806110 on Build GRCh38
rs886656263 Chr.9: 136806001-136806001 on Build GRCh38
rs200063481 Chr.9: 136806156-136806156 on Build GRCh38
rs200903563 Chr.9: 136806160-136806160 on Build GRCh38
rs1028633789 Chr.9: 136806202-136806202 on Build GRCh38
rs1030926097 Chr.9: 136806125-136806125 on Build GRCh38
rs774753826 Chr.9: 136806124-136806124 on Build GRCh38

View Full Product Details