Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 150389179-150389444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IYD
Entrez Gene ID
389434
Gene Name
iodotyrosine deiodinase
Gene Aliases
C6orf71, DEHAL1, IYD-1, TDH4
UniGene
Hs.310225
Gene Chromosome Location
Chr.6: 150368854-150404629 on Build GRCh38
Transcripts
NM_001164694.1 NM_001164695.1 NM_001318495.1 NM_203395.2 NR_134655.1 XM_006715478.3 XM_006715479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5807644 c.218A>G p.H73R Chr.6: 150389391-150389391 on Build GRCh38
4773048 c.238C>A p.H80N Chr.6: 150389411-150389411 on Build GRCh38
4693693 c.201A>C p.S67S Chr.6: 150389374-150389374 on Build GRCh38
4693688 c.182C>T p.A61V Chr.6: 150389355-150389355 on Build GRCh38
4532921 c.192G>A p.W64* Chr.6: 150389365-150389365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11963823 Chr.6: 150389270-150389270 on Build GRCh38
rs773408570 Chr.6: 150389326-150389327 on Build GRCh38
rs988016749 Chr.6: 150389222-150389222 on Build GRCh38
rs1016188994 Chr.6: 150389238-150389239 on Build GRCh38
rs1029195335 Chr.6: 150389215-150389215 on Build GRCh38
rs962463037 Chr.6: 150389256-150389256 on Build GRCh38
rs753195939 Chr.6: 150389294-150389294 on Build GRCh38
rs981233303 Chr.6: 150389295-150389295 on Build GRCh38
rs901996385 Chr.6: 150389208-150389208 on Build GRCh38
rs767202544 Chr.6: 150389318-150389318 on Build GRCh38
rs750084911 Chr.6: 150389349-150389349 on Build GRCh38
rs146414119 Chr.6: 150389415-150389415 on Build GRCh38
rs778042617 Chr.6: 150389296-150389314 on Build GRCh38
rs58734691 Chr.6: 150389279-150389279 on Build GRCh38
rs918452794 Chr.6: 150389293-150389293 on Build GRCh38
rs73617666 Chr.6: 150389412-150389412 on Build GRCh38
rs183806728 Chr.6: 150389249-150389249 on Build GRCh38
rs193293104 Chr.6: 150389217-150389217 on Build GRCh38
rs958130889 Chr.6: 150389211-150389211 on Build GRCh38
rs899437964 Chr.6: 150389357-150389357 on Build GRCh38
rs927202799 Chr.6: 150389299-150389299 on Build GRCh38
rs984762668 Chr.6: 150389226-150389226 on Build GRCh38
rs778799972 Chr.6: 150389390-150389390 on Build GRCh38
rs866634825 Chr.6: 150389365-150389365 on Build GRCh38
rs530589266 Chr.6: 150389240-150389240 on Build GRCh38
rs9397944 Chr.6: 150389311-150389311 on Build GRCh38
rs767219657 Chr.6: 150389414-150389414 on Build GRCh38
rs766062033 Chr.6: 150389369-150389369 on Build GRCh38
rs758471889 Chr.6: 150389413-150389413 on Build GRCh38
rs556323672 Chr.6: 150389234-150389234 on Build GRCh38
rs144193430 Chr.6: 150389354-150389354 on Build GRCh38
rs761374942 Chr.6: 150389317-150389317 on Build GRCh38
rs754825622 Chr.6: 150389388-150389388 on Build GRCh38
rs1054142316 Chr.6: 150389329-150389329 on Build GRCh38
rs753538030 Chr.6: 150389380-150389380 on Build GRCh38

View Full Product Details