Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11361102-11361363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPPR2
Entrez Gene ID
64748
Gene Name
phospholipid phosphatase related 2
Gene Aliases
LPPR2, PRG4
UniGene
Hs.6846
Gene Chromosome Location
Chr.19: 11355386-11365698 on Build GRCh38
Transcripts
NM_001170635.1 NM_022737.2 XM_005260017.1 XM_017027147.1 XM_017027148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223970 c.392G>A p.G131E Chr.19: 11361217-11361217 on Build GRCh38
4074129 c.429C>T p.F143F Chr.19: 11361254-11361254 on Build GRCh38
5461208 c.330C>T p.F110F Chr.19: 11361230-11361230 on Build GRCh38
32848 c.464C>T p.T155M Chr.19: 11361289-11361289 on Build GRCh38
4074131 c.467C>A p.P156Q Chr.19: 11361292-11361292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146450725 Chr.19: 11361219-11361219 on Build GRCh38
rs199627828 Chr.19: 11361305-11361305 on Build GRCh38
rs761258110 Chr.19: 11361284-11361284 on Build GRCh38
rs772537639 Chr.19: 11361239-11361241 on Build GRCh38
rs746925455 Chr.19: 11361196-11361196 on Build GRCh38
rs748704114 Chr.19: 11361191-11361191 on Build GRCh38
rs779138963 Chr.19: 11361182-11361182 on Build GRCh38
rs753875447 Chr.19: 11361308-11361308 on Build GRCh38
rs757425057 Chr.19: 11361180-11361180 on Build GRCh38
rs747640457 Chr.19: 11361210-11361210 on Build GRCh38
rs149837570 Chr.19: 11361156-11361156 on Build GRCh38
rs549113384 Chr.19: 11361339-11361339 on Build GRCh38
rs751291495 Chr.19: 11361334-11361334 on Build GRCh38
rs776199316 Chr.19: 11361278-11361278 on Build GRCh38
rs143046204 Chr.19: 11361314-11361314 on Build GRCh38
rs760019245 Chr.19: 11361247-11361247 on Build GRCh38
rs946749006 Chr.19: 11361166-11361166 on Build GRCh38
rs540988172 Chr.19: 11361165-11361165 on Build GRCh38
rs758146157 Chr.19: 11361193-11361193 on Build GRCh38
rs772544598 Chr.19: 11361263-11361263 on Build GRCh38
rs367608863 Chr.19: 11361321-11361321 on Build GRCh38
rs758235960 Chr.19: 11361325-11361325 on Build GRCh38
rs769358522 Chr.19: 11361212-11361212 on Build GRCh38
rs780004252 Chr.19: 11361194-11361194 on Build GRCh38
rs890312186 Chr.19: 11361274-11361274 on Build GRCh38
rs768224745 Chr.19: 11361204-11361204 on Build GRCh38
rs750237313 Chr.19: 11361184-11361184 on Build GRCh38
rs779808195 Chr.19: 11361330-11361330 on Build GRCh38
rs373730403 Chr.19: 11361320-11361320 on Build GRCh38
rs781138725 Chr.19: 11361346-11361346 on Build GRCh38
rs533767007 Chr.19: 11361143-11361143 on Build GRCh38

View Full Product Details