Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 261277-261550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234A
Entrez Gene ID
83986
Gene Name
family with sequence similarity 234 member A
Gene Aliases
C16orf9, ITFG3, gs19
UniGene
Hs.513225
Gene Chromosome Location
Chr.16: 234546-269943 on Build GRCh38
Transcripts
NM_001284497.1 NM_032039.3 NR_104317.1 XM_005255622.1 XM_005255623.2 XM_006720957.1 XM_011522691.1 XM_011522692.1 XM_017023760.1 XM_017023761.1 XM_017023762.1 XM_017023763.1 XM_017023764.1 XM_017023765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970143 c.660C>T p.D220D Chr.16: 261466-261466 on Build GRCh38
5178772 c.620C>T p.A207V Chr.16: 261426-261426 on Build GRCh38
6570011 c.637C>G p.L213V Chr.16: 261443-261443 on Build GRCh38
5178771 c.620C>T p.A207V Chr.16: 261426-261426 on Build GRCh38
4259788 c.631A>C p.S211R Chr.16: 261437-261437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771371860 Chr.16: 261530-261530 on Build GRCh38
rs772327712 Chr.16: 261423-261423 on Build GRCh38
rs375399489 Chr.16: 261467-261467 on Build GRCh38
rs531334759 Chr.16: 261318-261318 on Build GRCh38
rs922533282 Chr.16: 261472-261472 on Build GRCh38
rs141071695 Chr.16: 261362-261362 on Build GRCh38
rs757068037 Chr.16: 261412-261412 on Build GRCh38
rs566699080 Chr.16: 261328-261328 on Build GRCh38
rs370310669 Chr.16: 261349-261349 on Build GRCh38
rs768786381 Chr.16: 261419-261419 on Build GRCh38
rs764754338 Chr.16: 261390-261390 on Build GRCh38
rs748307992 Chr.16: 261485-261487 on Build GRCh38
rs544897570 Chr.16: 261485-261485 on Build GRCh38
rs985073107 Chr.16: 261297-261297 on Build GRCh38
rs910883955 Chr.16: 261527-261527 on Build GRCh38
rs556391006 Chr.16: 261462-261462 on Build GRCh38
rs757158347 Chr.16: 261470-261470 on Build GRCh38
rs371285767 Chr.16: 261367-261367 on Build GRCh38
rs910998485 Chr.16: 261327-261327 on Build GRCh38
rs549614841 Chr.16: 261320-261320 on Build GRCh38
rs200842636 Chr.16: 261480-261480 on Build GRCh38
rs779255204 Chr.16: 261503-261503 on Build GRCh38
rs771266398 Chr.16: 261427-261427 on Build GRCh38
rs757939754 Chr.16: 261391-261391 on Build GRCh38
rs759121877 Chr.16: 261374-261374 on Build GRCh38
rs759186759 Chr.16: 261435-261435 on Build GRCh38
rs560905982 Chr.16: 261317-261317 on Build GRCh38
rs774310414 Chr.16: 261356-261356 on Build GRCh38
rs74003712 Chr.16: 261368-261368 on Build GRCh38
rs551926447 Chr.16: 261326-261326 on Build GRCh38
rs761153670 Chr.16: 261426-261426 on Build GRCh38
rs186180742 Chr.16: 261496-261496 on Build GRCh38
rs758436154 Chr.16: 261514-261514 on Build GRCh38
rs73486203 Chr.16: 261507-261507 on Build GRCh38
rs180686936 Chr.16: 261469-261469 on Build GRCh38
rs768716498 Chr.16: 261354-261354 on Build GRCh38
rs376031368 Chr.16: 261357-261357 on Build GRCh38
rs779099999 Chr.16: 261420-261420 on Build GRCh38
rs371981131 Chr.16: 261404-261404 on Build GRCh38
rs376378664 Chr.16: 261486-261486 on Build GRCh38
rs967294690 Chr.16: 261477-261477 on Build GRCh38
rs376585702 Chr.16: 261531-261531 on Build GRCh38
rs777662192 Chr.16: 261400-261400 on Build GRCh38
rs748208304 Chr.16: 261421-261421 on Build GRCh38
rs764765725 Chr.16: 261449-261449 on Build GRCh38
rs773843743 Chr.16: 261424-261424 on Build GRCh38
rs539663540 Chr.16: 261324-261324 on Build GRCh38
rs754944213 Chr.16: 261340-261341 on Build GRCh38
rs573743054 Chr.16: 261417-261417 on Build GRCh38
rs375356087 Chr.16: 261418-261418 on Build GRCh38
rs367623020 Chr.16: 261466-261466 on Build GRCh38
rs772741018 Chr.16: 261361-261361 on Build GRCh38
rs747499504 Chr.16: 261509-261509 on Build GRCh38
rs571283074 Chr.16: 261323-261323 on Build GRCh38
rs538192068 Chr.16: 261431-261431 on Build GRCh38
rs781487565 Chr.16: 261349-261350 on Build GRCh38
rs749537312 Chr.16: 261351-261351 on Build GRCh38

View Full Product Details