Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27780291-27780538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL42
Entrez Gene ID
57542
Gene Name
kelch like family member 42
Gene Aliases
Ctb9, KLHDC5
UniGene
Hs.505104
Gene Chromosome Location
Chr.12: 27779957-27803040 on Build GRCh38
Transcripts
NM_020782.1 XM_017019698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500070 c.90C>T p.F30F Chr.12: 27780420-27780420 on Build GRCh38
6410698 c.18G>T p.M6I Chr.12: 27780348-27780348 on Build GRCh38
4792016 c.144C>A p.G48G Chr.12: 27780474-27780474 on Build GRCh38
6409411 c.90C>A p.F30L Chr.12: 27780420-27780420 on Build GRCh38
6376886 c.187C>T p.R63W Chr.12: 27780517-27780517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372461451 Chr.12: 27780442-27780442 on Build GRCh38
rs868826097 Chr.12: 27780380-27780380 on Build GRCh38
rs985158117 Chr.12: 27780361-27780361 on Build GRCh38
rs751036739 Chr.12: 27780414-27780414 on Build GRCh38
rs914327324 Chr.12: 27780322-27780322 on Build GRCh38
rs559230530 Chr.12: 27780465-27780465 on Build GRCh38
rs780622562 Chr.12: 27780451-27780451 on Build GRCh38
rs749872535 Chr.12: 27780312-27780312 on Build GRCh38
rs755549396 Chr.12: 27780459-27780459 on Build GRCh38
rs866345279 Chr.12: 27780433-27780433 on Build GRCh38
rs910868067 Chr.12: 27780363-27780363 on Build GRCh38
rs772244013 Chr.12: 27780346-27780346 on Build GRCh38
rs775288759 Chr.12: 27780377-27780377 on Build GRCh38
rs184964868 Chr.12: 27780426-27780426 on Build GRCh38
rs759271822 Chr.12: 27780368-27780368 on Build GRCh38
rs750103046 Chr.12: 27780438-27780438 on Build GRCh38
rs911425310 Chr.12: 27780416-27780416 on Build GRCh38
rs763009034 Chr.12: 27780378-27780378 on Build GRCh38
rs775390596 Chr.12: 27780505-27780505 on Build GRCh38
rs918702808 Chr.12: 27780447-27780447 on Build GRCh38
rs749735822 Chr.12: 27780455-27780455 on Build GRCh38
rs368325485 Chr.12: 27780322-27780322 on Build GRCh38
rs557821678 Chr.12: 27780471-27780471 on Build GRCh38
rs773446764 Chr.12: 27780366-27780366 on Build GRCh38
rs748287672 Chr.12: 27780323-27780323 on Build GRCh38
rs865849595 Chr.12: 27780405-27780405 on Build GRCh38
rs12820175 Chr.12: 27780458-27780458 on Build GRCh38
rs772154475 Chr.12: 27780470-27780470 on Build GRCh38
rs985719060 Chr.12: 27780370-27780370 on Build GRCh38
rs768601735 Chr.12: 27780518-27780518 on Build GRCh38
rs777908032 Chr.12: 27780476-27780476 on Build GRCh38
rs763476531 Chr.12: 27780408-27780408 on Build GRCh38
rs765023405 Chr.12: 27780372-27780372 on Build GRCh38
rs748211052 Chr.12: 27780469-27780469 on Build GRCh38
rs756540876 Chr.12: 27780439-27780439 on Build GRCh38
rs1006689036 Chr.12: 27780516-27780516 on Build GRCh38
rs12820185 Chr.12: 27780480-27780480 on Build GRCh38
rs369177330 Chr.12: 27780429-27780429 on Build GRCh38
rs769569067 Chr.12: 27780483-27780483 on Build GRCh38

View Full Product Details