Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18706821-18707080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
UniGene
Hs.434152
Gene Chromosome Location
Chr.11: 18704305-18726939 on Build GRCh38
Transcripts
NM_173588.3 XM_011520030.2 XM_011520031.2 XM_017017607.1 XM_017017608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369379 c.3454A>G p.N1152D Chr.11: 18707040-18707040 on Build GRCh38
5601627 c.3515G>A p.R1172Q Chr.11: 18706979-18706979 on Build GRCh38
6051546 c.3504A>T p.R1168S Chr.11: 18706990-18706990 on Build GRCh38
6367481 c.3448T>C p.F1150L Chr.11: 18707046-18707046 on Build GRCh38
4651080 c.3470C>T p.T1157M Chr.11: 18707024-18707024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765887771 Chr.11: 18706959-18706959 on Build GRCh38
rs957946008 Chr.11: 18707035-18707035 on Build GRCh38
rs761936333 Chr.11: 18706922-18706924 on Build GRCh38
rs773254114 Chr.11: 18707031-18707036 on Build GRCh38
rs771050426 Chr.11: 18706968-18706969 on Build GRCh38
rs1022544763 Chr.11: 18706941-18706941 on Build GRCh38
rs968612490 Chr.11: 18706971-18706971 on Build GRCh38
rs918781282 Chr.11: 18707061-18707061 on Build GRCh38
rs772377225 Chr.11: 18707005-18707006 on Build GRCh38
rs1056872660 Chr.11: 18706897-18706897 on Build GRCh38
rs910138453 Chr.11: 18706844-18706844 on Build GRCh38
rs117051022 Chr.11: 18707039-18707039 on Build GRCh38
rs978111691 Chr.11: 18707006-18707006 on Build GRCh38
rs977269627 Chr.11: 18707023-18707023 on Build GRCh38
rs369995667 Chr.11: 18707000-18707000 on Build GRCh38
rs897321807 Chr.11: 18706879-18706879 on Build GRCh38
rs564741325 Chr.11: 18707037-18707037 on Build GRCh38
rs772842457 Chr.11: 18706926-18706926 on Build GRCh38
rs967620782 Chr.11: 18707005-18707005 on Build GRCh38
rs758558229 Chr.11: 18707056-18707056 on Build GRCh38
rs868059119 Chr.11: 18706979-18706979 on Build GRCh38
rs767124973 Chr.11: 18707052-18707052 on Build GRCh38
rs533400398 Chr.11: 18706891-18706891 on Build GRCh38
rs922729846 Chr.11: 18707024-18707024 on Build GRCh38
rs750555153 Chr.11: 18706925-18706925 on Build GRCh38
rs1012553529 Chr.11: 18706930-18706930 on Build GRCh38
rs554822408 Chr.11: 18706875-18706875 on Build GRCh38
rs1019152146 Chr.11: 18707003-18707003 on Build GRCh38
rs12361825 Chr.11: 18706876-18706876 on Build GRCh38
rs941559818 Chr.11: 18706874-18706874 on Build GRCh38
rs955879407 Chr.11: 18707007-18707007 on Build GRCh38
rs762824940 Chr.11: 18706940-18706940 on Build GRCh38
rs909877046 Chr.11: 18707042-18707042 on Build GRCh38
rs543347161 Chr.11: 18707044-18707046 on Build GRCh38
rs941634954 Chr.11: 18707054-18707054 on Build GRCh38

View Full Product Details