Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127507872-127508143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129B
Entrez Gene ID
64855
Gene Name
family with sequence similarity 129 member B
Gene Aliases
C9orf88, MEG-3, MINERVA, OC58, bA356B19.6
UniGene
Hs.522401
Gene Chromosome Location
Chr.9: 127505338-127579007 on Build GRCh38
Transcripts
NM_001035534.2 NM_022833.3 XM_005252135.2 XM_011518925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472658 c.1607C>T p.T536M Chr.9: 127507914-127507914 on Build GRCh38
4467847 c.1510C>T p.L504L Chr.9: 127508125-127508125 on Build GRCh38
6867039 c.1512C>T p.P504P Chr.9: 127507970-127507970 on Build GRCh38
4467848 c.1471C>T p.L491L Chr.9: 127508125-127508125 on Build GRCh38
7096221 c.1460_1462delGCA p.S487delS Chr.9: 127508134-127508136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773426577 Chr.9: 127508084-127508084 on Build GRCh38
rs768695978 Chr.9: 127507993-127507993 on Build GRCh38
rs902306695 Chr.9: 127508054-127508054 on Build GRCh38
rs548522801 Chr.9: 127508022-127508022 on Build GRCh38
rs766780391 Chr.9: 127508021-127508021 on Build GRCh38
rs765640749 Chr.9: 127508110-127508110 on Build GRCh38
rs746590065 Chr.9: 127507963-127507963 on Build GRCh38
rs376456300 Chr.9: 127507913-127507913 on Build GRCh38
rs775295866 Chr.9: 127507985-127507985 on Build GRCh38
rs761004900 Chr.9: 127508016-127508016 on Build GRCh38
rs987789609 Chr.9: 127508013-127508013 on Build GRCh38
rs372002650 Chr.9: 127508061-127508061 on Build GRCh38
rs780940200 Chr.9: 127507969-127507969 on Build GRCh38
rs376674479 Chr.9: 127508120-127508120 on Build GRCh38
rs911442668 Chr.9: 127508046-127508046 on Build GRCh38
rs745642516 Chr.9: 127507971-127507971 on Build GRCh38
rs201427745 Chr.9: 127507968-127507968 on Build GRCh38
rs750783560 Chr.9: 127507914-127507914 on Build GRCh38
rs200735091 Chr.9: 127508074-127508074 on Build GRCh38
rs762901875 Chr.9: 127507900-127507900 on Build GRCh38
rs375909922 Chr.9: 127508010-127508010 on Build GRCh38
rs1023527621 Chr.9: 127507929-127507929 on Build GRCh38
rs61732274 Chr.9: 127507910-127507910 on Build GRCh38
rs370872769 Chr.9: 127508051-127508051 on Build GRCh38
rs764013369 Chr.9: 127507901-127507901 on Build GRCh38
rs759988005 Chr.9: 127508107-127508107 on Build GRCh38
rs779542946 Chr.9: 127508047-127508047 on Build GRCh38
rs893484306 Chr.9: 127508067-127508067 on Build GRCh38
rs769529372 Chr.9: 127507978-127507978 on Build GRCh38
rs985745694 Chr.9: 127508027-127508027 on Build GRCh38
rs200796169 Chr.9: 127508094-127508094 on Build GRCh38
rs374249421 Chr.9: 127508053-127508053 on Build GRCh38
rs750622226 Chr.9: 127508012-127508012 on Build GRCh38
rs375926749 Chr.9: 127508066-127508066 on Build GRCh38
rs777094883 Chr.9: 127507949-127507949 on Build GRCh38
rs780583830 Chr.9: 127507936-127507936 on Build GRCh38
rs761676573 Chr.9: 127508078-127508078 on Build GRCh38
rs761991205 Chr.9: 127508001-127508001 on Build GRCh38
rs780703594 Chr.9: 127508057-127508057 on Build GRCh38
rs568312636 Chr.9: 127508035-127508035 on Build GRCh38
rs755430904 Chr.9: 127508045-127508045 on Build GRCh38
rs778065232 Chr.9: 127508007-127508007 on Build GRCh38
rs774287879 Chr.9: 127507994-127507994 on Build GRCh38
rs981839202 Chr.9: 127507988-127507988 on Build GRCh38
rs754252948 Chr.9: 127508097-127508097 on Build GRCh38
rs774908788 Chr.9: 127508092-127508092 on Build GRCh38
rs199700640 Chr.9: 127507981-127507981 on Build GRCh38
rs749146441 Chr.9: 127508070-127508070 on Build GRCh38
rs751672920 Chr.9: 127507907-127507907 on Build GRCh38
rs373796684 Chr.9: 127507990-127507990 on Build GRCh38
rs755550771 Chr.9: 127507945-127507945 on Build GRCh38
rs529266916 Chr.9: 127507922-127507922 on Build GRCh38
rs760926345 Chr.9: 127508087-127508087 on Build GRCh38
rs772146953 Chr.9: 127508081-127508081 on Build GRCh38
rs769449421 Chr.9: 127508062-127508062 on Build GRCh38
rs749769768 Chr.9: 127507939-127507939 on Build GRCh38
rs550545019 Chr.9: 127508077-127508077 on Build GRCh38

View Full Product Details