Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179064152-179064395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20B
Entrez Gene ID
9917
Gene Name
family with sequence similarity 20 member B
Gene Aliases
gxk1
UniGene
Hs.5737
Gene Chromosome Location
Chr.1: 179025804-179076574 on Build GRCh38
Transcripts
NM_001324310.1 NM_001324311.1 NM_014864.3 XM_017003001.1 XM_017003002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74595 c.806G>T p.G269V Chr.1: 179064364-179064364 on Build GRCh38
6659409 c.835G>A p.A279T Chr.1: 179064393-179064393 on Build GRCh38
6362767 c.801C>A p.D267E Chr.1: 179064359-179064359 on Build GRCh38
1336628 c.811C>T p.R271C Chr.1: 179064369-179064369 on Build GRCh38
4824803 c.783G>A p.K261K Chr.1: 179064341-179064341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755919918 Chr.1: 179064273-179064273 on Build GRCh38
rs761046746 Chr.1: 179064320-179064320 on Build GRCh38
rs762613431 Chr.1: 179064329-179064329 on Build GRCh38
rs143013339 Chr.1: 179064371-179064371 on Build GRCh38
rs772629167 Chr.1: 179064294-179064294 on Build GRCh38
rs766660827 Chr.1: 179064325-179064325 on Build GRCh38
rs768815241 Chr.1: 179064289-179064289 on Build GRCh38
rs991571788 Chr.1: 179064276-179064276 on Build GRCh38
rs140406781 Chr.1: 179064366-179064366 on Build GRCh38
rs751715086 Chr.1: 179064270-179064271 on Build GRCh38
rs751034220 Chr.1: 179064333-179064333 on Build GRCh38
rs778279832 Chr.1: 179064370-179064370 on Build GRCh38
rs181895952 Chr.1: 179064181-179064181 on Build GRCh38
rs778968805 Chr.1: 179064367-179064367 on Build GRCh38
rs949837124 Chr.1: 179064290-179064290 on Build GRCh38
rs763781228 Chr.1: 179064332-179064332 on Build GRCh38
rs569672602 Chr.1: 179064217-179064217 on Build GRCh38
rs767476826 Chr.1: 179064347-179064347 on Build GRCh38
rs150381478 Chr.1: 179064368-179064368 on Build GRCh38
rs753736442 Chr.1: 179064362-179064362 on Build GRCh38
rs567995916 Chr.1: 179064356-179064356 on Build GRCh38
rs750341151 Chr.1: 179064348-179064348 on Build GRCh38
rs945691615 Chr.1: 179064245-179064245 on Build GRCh38
rs926881757 Chr.1: 179064211-179064211 on Build GRCh38
rs149038000 Chr.1: 179064252-179064252 on Build GRCh38
rs775086684 Chr.1: 179064326-179064326 on Build GRCh38
rs761292654 Chr.1: 179064346-179064346 on Build GRCh38
rs781027865 Chr.1: 179064255-179064255 on Build GRCh38
rs771280602 Chr.1: 179064373-179064373 on Build GRCh38
rs375106861 Chr.1: 179064287-179064287 on Build GRCh38
rs755870535 Chr.1: 179064351-179064351 on Build GRCh38
rs892828524 Chr.1: 179064218-179064218 on Build GRCh38
rs745667905 Chr.1: 179064266-179064266 on Build GRCh38
rs780025166 Chr.1: 179064355-179064355 on Build GRCh38
rs772062570 Chr.1: 179064369-179064369 on Build GRCh38
rs201538828 Chr.1: 179064336-179064336 on Build GRCh38
rs1036149063 Chr.1: 179064352-179064352 on Build GRCh38
rs773650162 Chr.1: 179064305-179064305 on Build GRCh38
rs372345743 Chr.1: 179064283-179064283 on Build GRCh38
rs1054109362 Chr.1: 179064199-179064199 on Build GRCh38

View Full Product Details