Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49641829-49642100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5051656 c.1753G>A p.V585I Chr.12: 49641959-49641959 on Build GRCh38
5049046 c.1765G>C p.D589H Chr.12: 49641971-49641971 on Build GRCh38
2008131 c.1716C>T p.C572C Chr.12: 49641922-49641922 on Build GRCh38
5646621 c.1752C>T p.H584H Chr.12: 49641958-49641958 on Build GRCh38
1222173 c.1717G>A p.V573M Chr.12: 49641923-49641923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768704025 Chr.12: 49641981-49641981 on Build GRCh38
rs572799556 Chr.12: 49641959-49641959 on Build GRCh38
rs552783251 Chr.12: 49641937-49641937 on Build GRCh38
rs758782882 Chr.12: 49641871-49641871 on Build GRCh38
rs143028418 Chr.12: 49641958-49641958 on Build GRCh38
rs763217658 Chr.12: 49642020-49642020 on Build GRCh38
rs200397242 Chr.12: 49641966-49641966 on Build GRCh38
rs746441634 Chr.12: 49641878-49641878 on Build GRCh38
rs767032476 Chr.12: 49642042-49642042 on Build GRCh38
rs371483209 Chr.12: 49642001-49642001 on Build GRCh38
rs138798728 Chr.12: 49641911-49641911 on Build GRCh38
rs948158296 Chr.12: 49641875-49641875 on Build GRCh38
rs749877244 Chr.12: 49642043-49642043 on Build GRCh38
rs922277973 Chr.12: 49642060-49642062 on Build GRCh38
rs139833340 Chr.12: 49641994-49641994 on Build GRCh38
rs756620011 Chr.12: 49641887-49641888 on Build GRCh38
rs577923928 Chr.12: 49641995-49641995 on Build GRCh38
rs764713565 Chr.12: 49642033-49642033 on Build GRCh38
rs926005426 Chr.12: 49641934-49641934 on Build GRCh38
rs368968591 Chr.12: 49641985-49641985 on Build GRCh38
rs916747450 Chr.12: 49641853-49641853 on Build GRCh38
rs768420141 Chr.12: 49641919-49641919 on Build GRCh38
rs778731129 Chr.12: 49642059-49642059 on Build GRCh38
rs752951274 Chr.12: 49641936-49641936 on Build GRCh38
rs1016658157 Chr.12: 49641976-49641976 on Build GRCh38
rs760879578 Chr.12: 49641986-49641986 on Build GRCh38
rs745546676 Chr.12: 49641963-49641963 on Build GRCh38
rs992411031 Chr.12: 49642046-49642046 on Build GRCh38
rs770639755 Chr.12: 49641883-49641883 on Build GRCh38
rs748968192 Chr.12: 49641975-49641975 on Build GRCh38
rs779567676 Chr.12: 49642044-49642044 on Build GRCh38
rs115713495 Chr.12: 49641912-49641912 on Build GRCh38
rs374723844 Chr.12: 49641956-49641956 on Build GRCh38
rs925288967 Chr.12: 49641890-49641890 on Build GRCh38
rs368121359 Chr.12: 49641872-49641872 on Build GRCh38
rs772898724 Chr.12: 49641908-49641908 on Build GRCh38
rs374992843 Chr.12: 49642038-49642038 on Build GRCh38
rs766700373 Chr.12: 49642054-49642054 on Build GRCh38
rs766758338 Chr.12: 49641923-49641923 on Build GRCh38
rs760830638 Chr.12: 49641922-49641922 on Build GRCh38
rs957897204 Chr.12: 49641953-49641953 on Build GRCh38
rs778142028 Chr.12: 49641952-49641952 on Build GRCh38
rs747146229 Chr.12: 49641893-49641893 on Build GRCh38
rs201024261 Chr.12: 49642039-49642039 on Build GRCh38
rs757718769 Chr.12: 49642041-49642041 on Build GRCh38
rs771890161 Chr.12: 49641867-49641867 on Build GRCh38
rs753248075 Chr.12: 49641881-49641882 on Build GRCh38
rs202101825 Chr.12: 49641889-49641889 on Build GRCh38
rs1043793356 Chr.12: 49641877-49641877 on Build GRCh38
rs1025312356 Chr.12: 49642007-49642007 on Build GRCh38
rs769383964 Chr.12: 49641964-49641964 on Build GRCh38
rs745708210 Chr.12: 49641894-49641894 on Build GRCh38
rs1024282470 Chr.12: 49641872-49641874 on Build GRCh38
rs542266861 Chr.12: 49641940-49641940 on Build GRCh38
rs759640650 Chr.12: 49641878-49641878 on Build GRCh38
rs771166750 Chr.12: 49641988-49641988 on Build GRCh38
rs542772866 Chr.12: 49641930-49641930 on Build GRCh38
rs767555159 Chr.12: 49641877-49641878 on Build GRCh38
rs749164218 Chr.12: 49642051-49642051 on Build GRCh38

View Full Product Details