Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152120467-152120732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAM2
Entrez Gene ID
10254
Gene Name
signal transducing adaptor molecule 2
Gene Aliases
Hbp, STAM2A, STAM2B
UniGene
Hs.17200
Gene Chromosome Location
Chr.2: 152116801-152175992 on Build GRCh38
Transcripts
NM_005843.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265372 c.1540C>A p.Q514K Chr.2: 152120612-152120612 on Build GRCh38
6587226 c.1550C>G p.T517R Chr.2: 152120602-152120602 on Build GRCh38
1161317 c.1561C>T p.Q521* Chr.2: 152120591-152120591 on Build GRCh38
6154108 c.1518A>T p.T506T Chr.2: 152120634-152120634 on Build GRCh38
5058158 c.1497A>G p.Q499Q Chr.2: 152120655-152120655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558350294 Chr.2: 152120691-152120691 on Build GRCh38
rs954214674 Chr.2: 152120621-152120621 on Build GRCh38
rs764341068 Chr.2: 152120577-152120577 on Build GRCh38
rs139852594 Chr.2: 152120562-152120562 on Build GRCh38
rs751819101 Chr.2: 152120647-152120647 on Build GRCh38
rs757728819 Chr.2: 152120539-152120539 on Build GRCh38
rs539537729 Chr.2: 152120564-152120564 on Build GRCh38
rs775117318 Chr.2: 152120600-152120600 on Build GRCh38
rs34505651 Chr.2: 152120524-152120525 on Build GRCh38
rs1029934052 Chr.2: 152120612-152120612 on Build GRCh38
rs16830707 Chr.2: 152120544-152120544 on Build GRCh38
rs986034837 Chr.2: 152120635-152120635 on Build GRCh38
rs200233333 Chr.2: 152120541-152120541 on Build GRCh38
rs752024988 Chr.2: 152120579-152120579 on Build GRCh38
rs747276193 Chr.2: 152120561-152120561 on Build GRCh38
rs776056405 Chr.2: 152120623-152120623 on Build GRCh38
rs200914607 Chr.2: 152120534-152120534 on Build GRCh38
rs202201725 Chr.2: 152120551-152120551 on Build GRCh38
rs902223118 Chr.2: 152120517-152120517 on Build GRCh38
rs748208971 Chr.2: 152120690-152120690 on Build GRCh38
rs991583213 Chr.2: 152120516-152120516 on Build GRCh38
rs369433659 Chr.2: 152120580-152120580 on Build GRCh38
rs765729847 Chr.2: 152120640-152120640 on Build GRCh38
rs149960401 Chr.2: 152120674-152120674 on Build GRCh38
rs763746100 Chr.2: 152120611-152120611 on Build GRCh38
rs763656500 Chr.2: 152120529-152120529 on Build GRCh38
rs771801034 Chr.2: 152120593-152120593 on Build GRCh38
rs781132076 Chr.2: 152120566-152120566 on Build GRCh38
rs977851744 Chr.2: 152120679-152120679 on Build GRCh38
rs774177131 Chr.2: 152120586-152120586 on Build GRCh38
rs766918705 Chr.2: 152120644-152120644 on Build GRCh38
rs549324409 Chr.2: 152120501-152120501 on Build GRCh38
rs779533787 Chr.2: 152120529-152120531 on Build GRCh38
rs780403657 Chr.2: 152120555-152120555 on Build GRCh38
rs761263937 Chr.2: 152120630-152120630 on Build GRCh38
rs910136977 Chr.2: 152120495-152120496 on Build GRCh38
rs201532688 Chr.2: 152120554-152120554 on Build GRCh38
rs368062610 Chr.2: 152120542-152120542 on Build GRCh38
rs148514064 Chr.2: 152120682-152120682 on Build GRCh38
rs750943315 Chr.2: 152120641-152120641 on Build GRCh38
rs760375468 Chr.2: 152120609-152120609 on Build GRCh38

View Full Product Details