Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67429468-67429741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KB2
Entrez Gene ID
6199
Gene Name
ribosomal protein S6 kinase B2
Gene Aliases
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
UniGene
Hs.534345
Gene Chromosome Location
Chr.11: 67428460-67435408 on Build GRCh38
Transcripts
NM_003952.2 XM_005274164.1 XM_005274165.4 XM_006718655.3 XM_006718656.3 XM_006718657.1 XM_017018108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5839374 c.265G>A p.G89S Chr.11: 67429551-67429551 on Build GRCh38
6976148 c.244T>C p.F82L Chr.11: 67429530-67429530 on Build GRCh38
6913562 c.281A>G p.K94R Chr.11: 67429567-67429567 on Build GRCh38
3359360 c.245T>G p.F82C Chr.11: 67429531-67429531 on Build GRCh38
6972966 c.270_278delCAACTTGGG p.N91_G93delNLG Chr.11: 67429556-67429564 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774712330 Chr.11: 67429536-67429536 on Build GRCh38
rs544066271 Chr.11: 67429617-67429617 on Build GRCh38
rs769862662 Chr.11: 67429635-67429635 on Build GRCh38
rs199520104 Chr.11: 67429613-67429613 on Build GRCh38
rs754495017 Chr.11: 67429570-67429570 on Build GRCh38
rs192512741 Chr.11: 67429688-67429688 on Build GRCh38
rs753225398 Chr.11: 67429557-67429557 on Build GRCh38
rs773641116 Chr.11: 67429643-67429643 on Build GRCh38
rs758095058 Chr.11: 67429589-67429589 on Build GRCh38
rs560288422 Chr.11: 67429689-67429689 on Build GRCh38
rs773737464 Chr.11: 67429540-67429540 on Build GRCh38
rs925793849 Chr.11: 67429674-67429674 on Build GRCh38
rs762169628 Chr.11: 67429538-67429538 on Build GRCh38
rs371886103 Chr.11: 67429602-67429602 on Build GRCh38
rs761051136 Chr.11: 67429543-67429543 on Build GRCh38
rs772459215 Chr.11: 67429641-67429641 on Build GRCh38
rs111404901 Chr.11: 67429652-67429652 on Build GRCh38
rs527426233 Chr.11: 67429701-67429701 on Build GRCh38
rs774624588 Chr.11: 67429628-67429628 on Build GRCh38
rs768677871 Chr.11: 67429527-67429527 on Build GRCh38
rs1004877931 Chr.11: 67429575-67429575 on Build GRCh38
rs746811418 Chr.11: 67429606-67429606 on Build GRCh38
rs764793251 Chr.11: 67429571-67429571 on Build GRCh38
rs532433397 Chr.11: 67429604-67429604 on Build GRCh38
rs201994522 Chr.11: 67429555-67429555 on Build GRCh38
rs989173786 Chr.11: 67429708-67429708 on Build GRCh38
rs749400034 Chr.11: 67429519-67429519 on Build GRCh38
rs978594498 Chr.11: 67429662-67429662 on Build GRCh38
rs752314615 Chr.11: 67429572-67429572 on Build GRCh38
rs965411413 Chr.11: 67429616-67429616 on Build GRCh38
rs780477249 Chr.11: 67429676-67429676 on Build GRCh38
rs376846654 Chr.11: 67429607-67429607 on Build GRCh38
rs781347554 Chr.11: 67429496-67429496 on Build GRCh38
rs770922755 Chr.11: 67429513-67429513 on Build GRCh38
rs367611886 Chr.11: 67429621-67429621 on Build GRCh38
rs749347874 Chr.11: 67429704-67429704 on Build GRCh38
rs143483478 Chr.11: 67429495-67429495 on Build GRCh38
rs370538354 Chr.11: 67429664-67429664 on Build GRCh38
rs748335158 Chr.11: 67429639-67429639 on Build GRCh38
rs374406780 Chr.11: 67429539-67429539 on Build GRCh38

View Full Product Details