Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98194749-98194974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPAL2
Entrez Gene ID
79815
Gene Name
NIPA like domain containing 2
Gene Aliases
NPAL2
UniGene
Hs.309489
Gene Chromosome Location
Chr.8: 98189826-98295427 on Build GRCh38
Transcripts
NM_001321635.1 NM_001321636.1 NM_024759.2 NR_135745.1 NR_135746.1 NR_135747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6837288 c.975G>C p.L325F Chr.8: 98194792-98194792 on Build GRCh38
6516403 c.952C>T p.L318L Chr.8: 98194815-98194815 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781555435 Chr.8: 98194835-98194835 on Build GRCh38
rs139183143 Chr.8: 98194806-98194806 on Build GRCh38
rs139680168 Chr.8: 98194900-98194900 on Build GRCh38
rs372334828 Chr.8: 98194889-98194889 on Build GRCh38
rs967693794 Chr.8: 98194811-98194811 on Build GRCh38
rs756329831 Chr.8: 98194845-98194845 on Build GRCh38
rs748307572 Chr.8: 98194842-98194842 on Build GRCh38
rs749440906 Chr.8: 98194847-98194847 on Build GRCh38
rs1048579283 Chr.8: 98194855-98194855 on Build GRCh38
rs755379377 Chr.8: 98194825-98194825 on Build GRCh38
rs766970350 Chr.8: 98194813-98194813 on Build GRCh38
rs190588686 Chr.8: 98194918-98194918 on Build GRCh38
rs763640709 Chr.8: 98194798-98194798 on Build GRCh38
rs1048256454 Chr.8: 98194910-98194910 on Build GRCh38
rs867046519 Chr.8: 98194779-98194779 on Build GRCh38
rs1014565442 Chr.8: 98194906-98194906 on Build GRCh38
rs967751249 Chr.8: 98194936-98194936 on Build GRCh38
rs185730364 Chr.8: 98194869-98194869 on Build GRCh38
rs774385017 Chr.8: 98194863-98194863 on Build GRCh38
rs753395360 Chr.8: 98194802-98194802 on Build GRCh38
rs756774222 Chr.8: 98194803-98194803 on Build GRCh38
rs367551918 Chr.8: 98194778-98194778 on Build GRCh38
rs760426043 Chr.8: 98194784-98194784 on Build GRCh38
rs751851260 Chr.8: 98194823-98194823 on Build GRCh38
rs3735889 Chr.8: 98194874-98194874 on Build GRCh38
rs895715136 Chr.8: 98194903-98194903 on Build GRCh38
rs770750550 Chr.8: 98194851-98194851 on Build GRCh38

View Full Product Details