Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27255125-27255398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL21
Entrez Gene ID
6144
Gene Name
ribosomal protein L21
Gene Aliases
HYPT12, L21
UniGene
Hs.381123
Gene Chromosome Location
Chr.13: 27251555-27256568 on Build GRCh38
Transcripts
NR_002574.1

Gene Symbol
SNORD102
Entrez Gene ID
26771
Gene Name
small nucleolar RNA, C/D box 102
Gene Aliases
RNU102, Z18
UniGene
Gene Chromosome Location
Chr.13: 27255064-27255135 on Build GRCh38
Transcripts
NR_002574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3468026 c.240T>C p.V80V Chr.13: 27255352-27255352 on Build GRCh38
3356387 c.204C>T p.T68T Chr.13: 27255316-27255316 on Build GRCh38
946407 c.148A>C p.K50Q Chr.13: 27255260-27255260 on Build GRCh38
2069956 c.235C>G p.Q79E Chr.13: 27255347-27255347 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371707085 Chr.13: 27255203-27255203 on Build GRCh38
rs753780264 Chr.13: 27255193-27255193 on Build GRCh38
rs749600972 Chr.13: 27255347-27255347 on Build GRCh38
rs757969429 Chr.13: 27255176-27255176 on Build GRCh38
rs780232683 Chr.13: 27255219-27255219 on Build GRCh38
rs756821526 Chr.13: 27255205-27255205 on Build GRCh38
rs766975338 Chr.13: 27255316-27255316 on Build GRCh38
rs775120705 Chr.13: 27255306-27255306 on Build GRCh38
rs200767103 Chr.13: 27255317-27255317 on Build GRCh38
rs756676608 Chr.13: 27255215-27255215 on Build GRCh38
rs756699442 Chr.13: 27255200-27255200 on Build GRCh38
rs759493455 Chr.13: 27255266-27255266 on Build GRCh38
rs902317594 Chr.13: 27255371-27255371 on Build GRCh38
rs756556213 Chr.13: 27255340-27255340 on Build GRCh38
rs763851131 Chr.13: 27255182-27255182 on Build GRCh38
rs965531441 Chr.13: 27255170-27255170 on Build GRCh38
rs748392465 Chr.13: 27255236-27255236 on Build GRCh38
rs371660516 Chr.13: 27255229-27255229 on Build GRCh38
rs768755560 Chr.13: 27255228-27255228 on Build GRCh38
rs773453390 Chr.13: 27255246-27255246 on Build GRCh38
rs763643191 Chr.13: 27255311-27255311 on Build GRCh38
rs779421826 Chr.13: 27255367-27255367 on Build GRCh38
rs747129293 Chr.13: 27255376-27255376 on Build GRCh38
rs367838328 Chr.13: 27255213-27255213 on Build GRCh38
rs950930768 Chr.13: 27255207-27255207 on Build GRCh38
rs61738475 Chr.13: 27255271-27255271 on Build GRCh38
rs1026444697 Chr.13: 27255190-27255190 on Build GRCh38
rs780695726 Chr.13: 27255206-27255206 on Build GRCh38
rs756896520 Chr.13: 27255181-27255181 on Build GRCh38
rs772209039 Chr.13: 27255244-27255244 on Build GRCh38
rs375524561 Chr.13: 27255309-27255309 on Build GRCh38
rs749958971 Chr.13: 27255321-27255321 on Build GRCh38
rs79811164 Chr.13: 27255314-27255314 on Build GRCh38
rs943343872 Chr.13: 27255257-27255257 on Build GRCh38
rs1042305895 Chr.13: 27255344-27255344 on Build GRCh38
rs772442788 Chr.13: 27255373-27255373 on Build GRCh38
rs987931607 Chr.13: 27255225-27255225 on Build GRCh38
rs751068625 Chr.13: 27255199-27255199 on Build GRCh38
rs780676823 Chr.13: 27255346-27255346 on Build GRCh38
rs748337276 Chr.13: 27255368-27255368 on Build GRCh38
rs56301318 Chr.13: 27255196-27255196 on Build GRCh38
rs923696408 Chr.13: 27255175-27255175 on Build GRCh38
rs914943436 Chr.13: 27255269-27255269 on Build GRCh38
rs763981107 Chr.13: 27255152-27255152 on Build GRCh38
rs3094293 Chr.13: 27255173-27255173 on Build GRCh38
rs749701636 Chr.13: 27255227-27255227 on Build GRCh38
rs552172668 Chr.13: 27255366-27255366 on Build GRCh38
rs752555371 Chr.13: 27255174-27255174 on Build GRCh38

View Full Product Details