Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68157798-68158023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFATC3
Entrez Gene ID
4775
Gene Name
nuclear factor of activated T-cells 3
Gene Aliases
NFAT4, NFATX
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68085366-68229259 on Build GRCh38
Transcripts
NM_004555.3 NM_173163.2 NM_173165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5628907 c.1432C>T p.L478L Chr.16: 68157899-68157899 on Build GRCh38
5612854 c.1416C>T p.N472N Chr.16: 68157883-68157883 on Build GRCh38
6725362 c.1481C>T p.A494V Chr.16: 68157948-68157948 on Build GRCh38
5628908 c.1432C>T p.L478L Chr.16: 68157899-68157899 on Build GRCh38
972678 c.1471C>T p.R491* Chr.16: 68157938-68157938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766619763 Chr.16: 68157990-68157990 on Build GRCh38
rs762077115 Chr.16: 68157963-68157963 on Build GRCh38
rs773543550 Chr.16: 68157925-68157925 on Build GRCh38
rs373061240 Chr.16: 68157832-68157832 on Build GRCh38
rs770818585 Chr.16: 68157911-68157911 on Build GRCh38
rs372761732 Chr.16: 68157985-68157985 on Build GRCh38
rs986342072 Chr.16: 68157848-68157848 on Build GRCh38
rs769592493 Chr.16: 68157921-68157921 on Build GRCh38
rs554478579 Chr.16: 68157943-68157943 on Build GRCh38
rs949395717 Chr.16: 68157918-68157918 on Build GRCh38
rs760841757 Chr.16: 68157986-68157986 on Build GRCh38
rs764726212 Chr.16: 68157836-68157836 on Build GRCh38
rs367576376 Chr.16: 68157982-68157982 on Build GRCh38
rs750727514 Chr.16: 68157882-68157882 on Build GRCh38
rs763504144 Chr.16: 68157834-68157834 on Build GRCh38
rs780852093 Chr.16: 68157912-68157912 on Build GRCh38
rs769362940 Chr.16: 68157829-68157829 on Build GRCh38
rs774977244 Chr.16: 68157959-68157959 on Build GRCh38
rs1007276029 Chr.16: 68157880-68157880 on Build GRCh38
rs762588316 Chr.16: 68157930-68157930 on Build GRCh38
rs756335867 Chr.16: 68157883-68157883 on Build GRCh38
rs200068848 Chr.16: 68157879-68157879 on Build GRCh38
rs150423397 Chr.16: 68157893-68157893 on Build GRCh38
rs780055283 Chr.16: 68157889-68157889 on Build GRCh38
rs753884452 Chr.16: 68157992-68157992 on Build GRCh38
rs777806475 Chr.16: 68157897-68157897 on Build GRCh38
rs376542866 Chr.16: 68157920-68157920 on Build GRCh38
rs746760486 Chr.16: 68157910-68157910 on Build GRCh38
rs574944220 Chr.16: 68157844-68157844 on Build GRCh38
rs762284805 Chr.16: 68157854-68157854 on Build GRCh38
rs753939817 Chr.16: 68157890-68157890 on Build GRCh38
rs368007528 Chr.16: 68157833-68157833 on Build GRCh38
rs767618382 Chr.16: 68157860-68157860 on Build GRCh38
rs775098782 Chr.16: 68157922-68157922 on Build GRCh38

View Full Product Details