Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79940714-79940969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D16
Entrez Gene ID
125058
Gene Name
TBC1 domain family member 16
Gene Aliases
-
UniGene
Hs.369819
Gene Chromosome Location
Chr.17: 79932343-80035875 on Build GRCh38
Transcripts
NM_001271844.1 NM_001271845.1 NM_019020.3 XM_005257049.2 XM_005257050.3 XM_006721694.3 XM_011524322.2 XM_011524323.2 XM_011524324.2 XM_017024186.1 XM_017024187.1 XM_017024188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370953 c.2246T>C p.L749P Chr.17: 79940917-79940917 on Build GRCh38
5055838 c.2276C>T p.P759L Chr.17: 79940887-79940887 on Build GRCh38
1386970 c.2283C>T p.D761D Chr.17: 79940880-79940880 on Build GRCh38
6797225 c.2211C>T p.Y737Y Chr.17: 79940952-79940952 on Build GRCh38
708775 c.2199G>A p.E733E Chr.17: 79940964-79940964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534526454 Chr.17: 79940858-79940858 on Build GRCh38
rs187446235 Chr.17: 79940908-79940908 on Build GRCh38
rs990560755 Chr.17: 79940744-79940744 on Build GRCh38
rs893923350 Chr.17: 79940842-79940842 on Build GRCh38
rs562826422 Chr.17: 79940810-79940810 on Build GRCh38
rs149758622 Chr.17: 79940898-79940898 on Build GRCh38
rs776926942 Chr.17: 79940926-79940926 on Build GRCh38
rs542474724 Chr.17: 79940746-79940746 on Build GRCh38
rs776054508 Chr.17: 79940856-79940856 on Build GRCh38
rs200852370 Chr.17: 79940880-79940880 on Build GRCh38
rs893869739 Chr.17: 79940907-79940907 on Build GRCh38
rs1009852729 Chr.17: 79940791-79940791 on Build GRCh38
rs548519011 Chr.17: 79940847-79940847 on Build GRCh38
rs781092548 Chr.17: 79940886-79940886 on Build GRCh38
rs765262608 Chr.17: 79940809-79940809 on Build GRCh38
rs566675159 Chr.17: 79940850-79940850 on Build GRCh38
rs140818887 Chr.17: 79940866-79940866 on Build GRCh38
rs769037123 Chr.17: 79940860-79940860 on Build GRCh38
rs367785113 Chr.17: 79940873-79940873 on Build GRCh38
rs62076647 Chr.17: 79940763-79940763 on Build GRCh38
rs759469164 Chr.17: 79940951-79940951 on Build GRCh38
rs769600969 Chr.17: 79940849-79940849 on Build GRCh38
rs765462588 Chr.17: 79940867-79940867 on Build GRCh38
rs368601528 Chr.17: 79940891-79940891 on Build GRCh38
rs770454634 Chr.17: 79940901-79940901 on Build GRCh38
rs747770269 Chr.17: 79940848-79940848 on Build GRCh38
rs1004161684 Chr.17: 79940925-79940925 on Build GRCh38
rs762166679 Chr.17: 79940927-79940927 on Build GRCh38
rs774324361 Chr.17: 79940948-79940948 on Build GRCh38
rs1016679782 Chr.17: 79940872-79940872 on Build GRCh38
rs773468374 Chr.17: 79940943-79940943 on Build GRCh38
rs147064065 Chr.17: 79940764-79940764 on Build GRCh38
rs764742864 Chr.17: 79940809-79940809 on Build GRCh38
rs959024027 Chr.17: 79940861-79940861 on Build GRCh38
rs769177756 Chr.17: 79940917-79940917 on Build GRCh38
rs751513582 Chr.17: 79940841-79940841 on Build GRCh38
rs906790302 Chr.17: 79940923-79940923 on Build GRCh38
rs777563835 Chr.17: 79940890-79940890 on Build GRCh38
rs1007557094 Chr.17: 79940837-79940837 on Build GRCh38
rs565318068 Chr.17: 79940782-79940782 on Build GRCh38
rs374588775 Chr.17: 79940853-79940853 on Build GRCh38
rs766450766 Chr.17: 79940947-79940947 on Build GRCh38
rs780042267 Chr.17: 79940833-79940833 on Build GRCh38
rs374603858 Chr.17: 79940887-79940887 on Build GRCh38
rs770415246 Chr.17: 79940855-79940855 on Build GRCh38
rs1296379 Chr.17: 79940844-79940844 on Build GRCh38
rs763080250 Chr.17: 79940874-79940874 on Build GRCh38
rs770206460 Chr.17: 79940942-79940942 on Build GRCh38
rs112102620 Chr.17: 79940889-79940889 on Build GRCh38
rs551079263 Chr.17: 79940802-79940802 on Build GRCh38
rs35133617 Chr.17: 79940938-79940939 on Build GRCh38
rs182976679 Chr.17: 79940800-79940800 on Build GRCh38
rs986725651 Chr.17: 79940885-79940885 on Build GRCh38
rs2241950 Chr.17: 79940762-79940762 on Build GRCh38
rs149141506 Chr.17: 79940944-79940944 on Build GRCh38
rs201904103 Chr.17: 79940879-79940879 on Build GRCh38
rs777014672 Chr.17: 79940862-79940862 on Build GRCh38

View Full Product Details