Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 145999989-146000234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3RF2
Entrez Gene ID
153769
Gene Name
SH3 domain containing ring finger 2
Gene Aliases
HEPP1, POSHER, PPP1R39, RNF158
UniGene
Hs.443728
Gene Chromosome Location
Chr.5: 145936563-146081773 on Build GRCh38
Transcripts
NM_152550.3 XM_005268380.2 XM_006714757.1 XM_011537567.2 XM_011537568.2 XM_011537569.2 XM_017009136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754091 c.478G>A p.E160K Chr.5: 146000157-146000157 on Build GRCh38
1434864 c.464G>A p.R155Q Chr.5: 146000143-146000143 on Build GRCh38
5654744 c.388G>C p.A130P Chr.5: 146000067-146000067 on Build GRCh38
5627923 c.385C>T p.R129* Chr.5: 146000064-146000064 on Build GRCh38
3611951 c.546C>T p.I182I Chr.5: 146000225-146000225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918464477 Chr.5: 146000172-146000172 on Build GRCh38
rs999477433 Chr.5: 146000030-146000030 on Build GRCh38
rs748682008 Chr.5: 146000121-146000121 on Build GRCh38
rs943729513 Chr.5: 146000212-146000212 on Build GRCh38
rs371869103 Chr.5: 146000142-146000142 on Build GRCh38
rs763952272 Chr.5: 146000079-146000079 on Build GRCh38
rs182905248 Chr.5: 146000162-146000162 on Build GRCh38
rs960705038 Chr.5: 146000124-146000124 on Build GRCh38
rs192447564 Chr.5: 146000210-146000210 on Build GRCh38
rs754408250 Chr.5: 146000174-146000174 on Build GRCh38
rs750506095 Chr.5: 146000179-146000179 on Build GRCh38
rs780317344 Chr.5: 146000191-146000191 on Build GRCh38
rs778480634 Chr.5: 146000064-146000064 on Build GRCh38
rs753961992 Chr.5: 146000065-146000065 on Build GRCh38
rs188024809 Chr.5: 146000192-146000192 on Build GRCh38
rs200412759 Chr.5: 146000163-146000163 on Build GRCh38
rs770489639 Chr.5: 146000137-146000137 on Build GRCh38
rs778941558 Chr.5: 146000089-146000089 on Build GRCh38
rs201618817 Chr.5: 146000025-146000025 on Build GRCh38
rs775984140 Chr.5: 146000157-146000157 on Build GRCh38
rs759763537 Chr.5: 146000031-146000031 on Build GRCh38
rs767947893 Chr.5: 146000037-146000037 on Build GRCh38
rs755910752 Chr.5: 146000201-146000201 on Build GRCh38
rs138522916 Chr.5: 146000131-146000131 on Build GRCh38
rs757233835 Chr.5: 146000175-146000175 on Build GRCh38
rs367562446 Chr.5: 146000114-146000114 on Build GRCh38
rs376743854 Chr.5: 146000103-146000103 on Build GRCh38
rs757434792 Chr.5: 146000208-146000208 on Build GRCh38
rs544952649 Chr.5: 146000213-146000213 on Build GRCh38
rs749992846 Chr.5: 146000021-146000021 on Build GRCh38
rs765386269 Chr.5: 146000176-146000176 on Build GRCh38
rs767319075 Chr.5: 146000154-146000154 on Build GRCh38
rs991246881 Chr.5: 146000133-146000133 on Build GRCh38
rs372705391 Chr.5: 146000042-146000042 on Build GRCh38
rs757360388 Chr.5: 146000077-146000077 on Build GRCh38
rs137960488 Chr.5: 146000189-146000189 on Build GRCh38
rs751617770 Chr.5: 146000170-146000170 on Build GRCh38
rs756667272 Chr.5: 146000045-146000045 on Build GRCh38
rs34942619 Chr.5: 146000199-146000199 on Build GRCh38
rs769709994 Chr.5: 146000119-146000119 on Build GRCh38
rs754685731 Chr.5: 146000102-146000102 on Build GRCh38
rs777631909 Chr.5: 146000207-146000207 on Build GRCh38
rs140312306 Chr.5: 146000143-146000143 on Build GRCh38
rs746121689 Chr.5: 146000092-146000092 on Build GRCh38

View Full Product Details