Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186301531-186301760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5350017 c.205T>C p.S69P Chr.1: 186301597-186301597 on Build GRCh38
6060376 c.292C>G p.P98A Chr.1: 186301684-186301684 on Build GRCh38
3418513 c.309C>A p.F103L Chr.1: 186301701-186301701 on Build GRCh38
167091 c.234C>T p.F78F Chr.1: 186301626-186301626 on Build GRCh38
1167893 c.235G>A p.E79K Chr.1: 186301627-186301627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776305395 Chr.1: 186301601-186301601 on Build GRCh38
rs757314301 Chr.1: 186301684-186301684 on Build GRCh38
rs780120758 Chr.1: 186301736-186301736 on Build GRCh38
rs767559777 Chr.1: 186301651-186301651 on Build GRCh38
rs768957112 Chr.1: 186301700-186301700 on Build GRCh38
rs201230692 Chr.1: 186301574-186301574 on Build GRCh38
rs142890978 Chr.1: 186301708-186301708 on Build GRCh38
rs775909741 Chr.1: 186301621-186301621 on Build GRCh38
rs371998874 Chr.1: 186301554-186301554 on Build GRCh38
rs750539223 Chr.1: 186301653-186301653 on Build GRCh38
rs746951033 Chr.1: 186301577-186301577 on Build GRCh38
rs375207109 Chr.1: 186301579-186301579 on Build GRCh38
rs746896075 Chr.1: 186301674-186301674 on Build GRCh38
rs369121158 Chr.1: 186301669-186301669 on Build GRCh38
rs774798475 Chr.1: 186301644-186301644 on Build GRCh38
rs963508234 Chr.1: 186301613-186301613 on Build GRCh38
rs1031162786 Chr.1: 186301582-186301582 on Build GRCh38
rs369769286 Chr.1: 186301654-186301654 on Build GRCh38
rs149854800 Chr.1: 186301737-186301737 on Build GRCh38
rs769360017 Chr.1: 186301687-186301687 on Build GRCh38
rs764445797 Chr.1: 186301642-186301642 on Build GRCh38
rs116639905 Chr.1: 186301668-186301668 on Build GRCh38
rs748999850 Chr.1: 186301693-186301693 on Build GRCh38
rs763169281 Chr.1: 186301641-186301641 on Build GRCh38
rs772036802 Chr.1: 186301569-186301569 on Build GRCh38
rs747964562 Chr.1: 186301566-186301566 on Build GRCh38
rs759205502 Chr.1: 186301609-186301609 on Build GRCh38
rs774938540 Chr.1: 186301691-186301691 on Build GRCh38
rs780818809 Chr.1: 186301685-186301685 on Build GRCh38
rs769372758 Chr.1: 186301610-186301610 on Build GRCh38
rs753899754 Chr.1: 186301723-186301723 on Build GRCh38
rs745437002 Chr.1: 186301686-186301686 on Build GRCh38
rs766571266 Chr.1: 186301658-186301658 on Build GRCh38
rs192682991 Chr.1: 186301724-186301724 on Build GRCh38
rs1038189180 Chr.1: 186301726-186301726 on Build GRCh38
rs144933686 Chr.1: 186301659-186301659 on Build GRCh38
rs768075373 Chr.1: 186301721-186301721 on Build GRCh38
rs955798670 Chr.1: 186301593-186301593 on Build GRCh38
rs762328095 Chr.1: 186301650-186301650 on Build GRCh38
rs762273156 Chr.1: 186301719-186301719 on Build GRCh38
rs985105750 Chr.1: 186301710-186301710 on Build GRCh38

View Full Product Details