Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21161733-21161992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKA3
Entrez Gene ID
221150
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
C13orf3, RAMA1
UniGene
Hs.88523
Gene Chromosome Location
Chr.13: 21153595-21176602 on Build GRCh38
Transcripts
NM_001166017.1 NM_145061.5 XM_005266288.3 XM_011534994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
147641 c.760A>G p.T254A Chr.13: 21161859-21161859 on Build GRCh38
4046456 c.751G>A p.A251T Chr.13: 21161868-21161868 on Build GRCh38
5376507 c.810C>T p.I270I Chr.13: 21161809-21161809 on Build GRCh38
4778210 c.760A>G p.T254A Chr.13: 21161859-21161859 on Build GRCh38
1300068 c.757G>A p.D253N Chr.13: 21161862-21161862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778222028 Chr.13: 21161809-21161809 on Build GRCh38
rs771399461 Chr.13: 21161827-21161827 on Build GRCh38
rs569387010 Chr.13: 21161861-21161861 on Build GRCh38
rs1040894850 Chr.13: 21161791-21161791 on Build GRCh38
rs747813209 Chr.13: 21161814-21161814 on Build GRCh38
rs764149474 Chr.13: 21161901-21161901 on Build GRCh38
rs200945102 Chr.13: 21161878-21161878 on Build GRCh38
rs776792211 Chr.13: 21161881-21161881 on Build GRCh38
rs993543737 Chr.13: 21161850-21161850 on Build GRCh38
rs776032418 Chr.13: 21161905-21161906 on Build GRCh38
rs778493859 Chr.13: 21161878-21161884 on Build GRCh38
rs771798304 Chr.13: 21161820-21161820 on Build GRCh38
rs140935640 Chr.13: 21161858-21161858 on Build GRCh38
rs759954172 Chr.13: 21161843-21161843 on Build GRCh38
rs760112450 Chr.13: 21161882-21161882 on Build GRCh38
rs763071505 Chr.13: 21161888-21161888 on Build GRCh38
rs746228168 Chr.13: 21161878-21161879 on Build GRCh38
rs377144863 Chr.13: 21161763-21161763 on Build GRCh38
rs202186176 Chr.13: 21161879-21161879 on Build GRCh38
rs752673165 Chr.13: 21161851-21161851 on Build GRCh38
rs943372796 Chr.13: 21161790-21161790 on Build GRCh38
rs868854122 Chr.13: 21161828-21161828 on Build GRCh38
rs533128475 Chr.13: 21161950-21161950 on Build GRCh38
rs777253375 Chr.13: 21161841-21161841 on Build GRCh38
rs17345690 Chr.13: 21161859-21161859 on Build GRCh38
rs765361685 Chr.13: 21161849-21161849 on Build GRCh38
rs370648548 Chr.13: 21161772-21161772 on Build GRCh38
rs761158054 Chr.13: 21161823-21161823 on Build GRCh38
rs901928718 Chr.13: 21161838-21161838 on Build GRCh38
rs773739283 Chr.13: 21161822-21161822 on Build GRCh38
rs755873621 Chr.13: 21161769-21161769 on Build GRCh38
rs770162085 Chr.13: 21161883-21161883 on Build GRCh38
rs202104082 Chr.13: 21161789-21161789 on Build GRCh38
rs764341389 Chr.13: 21161857-21161857 on Build GRCh38
rs775945779 Chr.13: 21161887-21161887 on Build GRCh38
rs991162343 Chr.13: 21161964-21161964 on Build GRCh38
rs754810578 Chr.13: 21161862-21161862 on Build GRCh38
rs749172673 Chr.13: 21161784-21161784 on Build GRCh38
rs751850273 Chr.13: 21161924-21161924 on Build GRCh38
rs1029552339 Chr.13: 21161918-21161918 on Build GRCh38
rs772369164 Chr.13: 21161881-21161881 on Build GRCh38
rs768281032 Chr.13: 21161802-21161802 on Build GRCh38
rs1040375472 Chr.13: 21161864-21161864 on Build GRCh38
rs778504634 Chr.13: 21161871-21161871 on Build GRCh38
rs200597155 Chr.13: 21161876-21161876 on Build GRCh38

View Full Product Details