Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119102187-119102440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRRM4
Entrez Gene ID
84530
Gene Name
serine/arginine repetitive matrix 4
Gene Aliases
KIAA1853, MU-MB-2.76, nSR100
UniGene
Hs.744964
Gene Chromosome Location
Chr.12: 118981495-119163051 on Build GRCh38
Transcripts
NM_194286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227154 c.559A>T p.R187* Chr.12: 119102372-119102372 on Build GRCh38
6569040 c.527G>C p.R176T Chr.12: 119102340-119102340 on Build GRCh38
1240325 c.424A>G p.T142A Chr.12: 119102237-119102237 on Build GRCh38
5909304 c.251G>A p.R84Q Chr.12: 119102355-119102355 on Build GRCh38
4835375 c.250C>G p.R84G Chr.12: 119102354-119102354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770430561 Chr.12: 119102217-119102217 on Build GRCh38
rs73213729 Chr.12: 119102257-119102257 on Build GRCh38
rs375971257 Chr.12: 119102372-119102372 on Build GRCh38
rs543604828 Chr.12: 119102322-119102322 on Build GRCh38
rs758138809 Chr.12: 119102278-119102278 on Build GRCh38
rs780825848 Chr.12: 119102400-119102400 on Build GRCh38
rs182742763 Chr.12: 119102244-119102244 on Build GRCh38
rs759671607 Chr.12: 119102245-119102245 on Build GRCh38
rs779345328 Chr.12: 119102314-119102314 on Build GRCh38
rs116273476 Chr.12: 119102401-119102401 on Build GRCh38
rs574984677 Chr.12: 119102303-119102303 on Build GRCh38
rs373924054 Chr.12: 119102311-119102311 on Build GRCh38
rs761868587 Chr.12: 119102350-119102350 on Build GRCh38
rs775549365 Chr.12: 119102255-119102255 on Build GRCh38
rs371343772 Chr.12: 119102329-119102329 on Build GRCh38
rs765147050 Chr.12: 119102271-119102271 on Build GRCh38
rs137921110 Chr.12: 119102333-119102333 on Build GRCh38
rs368148077 Chr.12: 119102312-119102312 on Build GRCh38
rs377246079 Chr.12: 119102407-119102407 on Build GRCh38
rs754510647 Chr.12: 119102399-119102399 on Build GRCh38
rs746251055 Chr.12: 119102409-119102409 on Build GRCh38
rs1041875933 Chr.12: 119102254-119102254 on Build GRCh38
rs371633893 Chr.12: 119102248-119102248 on Build GRCh38
rs545899062 Chr.12: 119102397-119102397 on Build GRCh38
rs773674182 Chr.12: 119102219-119102219 on Build GRCh38
rs746008357 Chr.12: 119102324-119102324 on Build GRCh38
rs118023042 Chr.12: 119102258-119102258 on Build GRCh38
rs908392170 Chr.12: 119102371-119102371 on Build GRCh38
rs771307455 Chr.12: 119102307-119102348 on Build GRCh38
rs750031037 Chr.12: 119102277-119102277 on Build GRCh38
rs150353576 Chr.12: 119102331-119102331 on Build GRCh38
rs1011723928 Chr.12: 119102246-119102246 on Build GRCh38
rs776426944 Chr.12: 119102344-119102344 on Build GRCh38
rs757783698 Chr.12: 119102403-119102403 on Build GRCh38
rs73213730 Chr.12: 119102398-119102398 on Build GRCh38
rs771380437 Chr.12: 119102242-119102242 on Build GRCh38
rs760719631 Chr.12: 119102332-119102332 on Build GRCh38
rs761117796 Chr.12: 119102355-119102355 on Build GRCh38
rs201474953 Chr.12: 119102304-119102304 on Build GRCh38
rs779640585 Chr.12: 119102281-119102281 on Build GRCh38
rs558491338 Chr.12: 119102224-119102224 on Build GRCh38
rs761482318 Chr.12: 119102268-119102268 on Build GRCh38
rs764884067 Chr.12: 119102354-119102354 on Build GRCh38

View Full Product Details