Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70062443-70062702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD6A
Entrez Gene ID
139562
Gene Name
OTU deubiquitinase 6A
Gene Aliases
DUBA2, HSHIN6
UniGene
Hs.447381
Gene Chromosome Location
Chr.X: 70062491-70064179 on Build GRCh38
Transcripts
NM_207320.2

SNP Detail

SNP ID Chromosome Location
rs769666862 Chr.X: 70062538-70062538 on Build GRCh38
rs1054630394 Chr.X: 70062682-70062682 on Build GRCh38
rs754168828 Chr.X: 70062632-70062632 on Build GRCh38
rs559111619 Chr.X: 70062641-70062641 on Build GRCh38
rs867118856 Chr.X: 70062673-70062673 on Build GRCh38
rs773523829 Chr.X: 70062561-70062561 on Build GRCh38
rs749281386 Chr.X: 70062540-70062540 on Build GRCh38
rs760055968 Chr.X: 70062633-70062633 on Build GRCh38
rs756927032 Chr.X: 70062534-70062534 on Build GRCh38
rs890879201 Chr.X: 70062631-70062631 on Build GRCh38
rs202156115 Chr.X: 70062543-70062543 on Build GRCh38
rs765873831 Chr.X: 70062640-70062640 on Build GRCh38
rs780917824 Chr.X: 70062535-70062535 on Build GRCh38
rs376752285 Chr.X: 70062683-70062683 on Build GRCh38
rs772262444 Chr.X: 70062552-70062552 on Build GRCh38
rs766829102 Chr.X: 70062603-70062603 on Build GRCh38
rs774347339 Chr.X: 70062548-70062548 on Build GRCh38
rs868819954 Chr.X: 70062623-70062623 on Build GRCh38
rs779850607 Chr.X: 70062539-70062539 on Build GRCh38
rs902969540 Chr.X: 70062575-70062575 on Build GRCh38
rs777363758 Chr.X: 70062476-70062476 on Build GRCh38
rs898191675 Chr.X: 70062466-70062466 on Build GRCh38
rs756871870 Chr.X: 70062666-70062666 on Build GRCh38
rs751121663 Chr.X: 70062515-70062515 on Build GRCh38
rs999913338 Chr.X: 70062658-70062658 on Build GRCh38
rs761904845 Chr.X: 70062549-70062549 on Build GRCh38
rs750959542 Chr.X: 70062536-70062536 on Build GRCh38
rs372088322 Chr.X: 70062604-70062604 on Build GRCh38
rs760944430 Chr.X: 70062573-70062573 on Build GRCh38
rs757973021 Chr.X: 70062475-70062475 on Build GRCh38

View Full Product Details