Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136792710-136792957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM141
Entrez Gene ID
85014
Gene Name
transmembrane protein 141
Gene Aliases
-
UniGene
Hs.356744
Gene Chromosome Location
Chr.9: 136791325-136793317 on Build GRCh38
Transcripts
NM_032928.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3699285 c.325T>C p.*109Q Chr.9: 136792830-136792830 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886070876 Chr.9: 136792881-136792881 on Build GRCh38
rs757769670 Chr.9: 136792837-136792837 on Build GRCh38
rs978766511 Chr.9: 136792910-136792910 on Build GRCh38
rs200782196 Chr.9: 136792866-136792866 on Build GRCh38
rs371577105 Chr.9: 136792778-136792778 on Build GRCh38
rs1016235958 Chr.9: 136792891-136792891 on Build GRCh38
rs763902345 Chr.9: 136792781-136792781 on Build GRCh38
rs775166355 Chr.9: 136792777-136792777 on Build GRCh38
rs769414754 Chr.9: 136792776-136792776 on Build GRCh38
rs368651425 Chr.9: 136792774-136792774 on Build GRCh38
rs370536028 Chr.9: 136792902-136792902 on Build GRCh38
rs202223156 Chr.9: 136792846-136792846 on Build GRCh38
rs771044861 Chr.9: 136792912-136792912 on Build GRCh38
rs755145111 Chr.9: 136792800-136792800 on Build GRCh38
rs1033013845 Chr.9: 136792925-136792925 on Build GRCh38
rs182473401 Chr.9: 136792761-136792761 on Build GRCh38
rs769255496 Chr.9: 136792852-136792852 on Build GRCh38
rs545385653 Chr.9: 136792899-136792899 on Build GRCh38
rs930353439 Chr.9: 136792865-136792865 on Build GRCh38
rs774032352 Chr.9: 136792785-136792785 on Build GRCh38
rs368907705 Chr.9: 136792863-136792863 on Build GRCh38
rs376572084 Chr.9: 136792893-136792893 on Build GRCh38
rs191276228 Chr.9: 136792840-136792840 on Build GRCh38
rs558094180 Chr.9: 136792737-136792737 on Build GRCh38
rs186370422 Chr.9: 136792792-136792792 on Build GRCh38
rs778132428 Chr.9: 136792828-136792828 on Build GRCh38
rs758560863 Chr.9: 136792821-136792821 on Build GRCh38
rs768201740 Chr.9: 136792876-136792876 on Build GRCh38
rs747400151 Chr.9: 136792833-136792833 on Build GRCh38
rs890701919 Chr.9: 136792729-136792729 on Build GRCh38
rs760592286 Chr.9: 136792805-136792806 on Build GRCh38
rs138348706 Chr.9: 136792818-136792818 on Build GRCh38
rs774195415 Chr.9: 136792882-136792882 on Build GRCh38
rs10485 Chr.9: 136792847-136792847 on Build GRCh38
rs998746658 Chr.9: 136792903-136792903 on Build GRCh38
rs1028434180 Chr.9: 136792911-136792911 on Build GRCh38
rs1004468898 Chr.9: 136792887-136792887 on Build GRCh38
rs761747554 Chr.9: 136792791-136792791 on Build GRCh38
rs1000248751 Chr.9: 136792918-136792918 on Build GRCh38
rs776113599 Chr.9: 136792769-136792769 on Build GRCh38
rs147664045 Chr.9: 136792762-136792762 on Build GRCh38

View Full Product Details