Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66668862-66669087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM4B
Entrez Gene ID
83759
Gene Name
RNA binding motif protein 4B
Gene Aliases
RBM30, RBM4L, ZCCHC15, ZCCHC21B, ZCRB3B
UniGene
Hs.618713
Gene Chromosome Location
Chr.11: 66664994-66677921 on Build GRCh38
Transcripts
NM_001286135.1 NM_031492.3 XM_011545297.2 XM_017018402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
289969 c.805_806insT p.D269fs*4 Chr.11: 66668898-66668899 on Build GRCh38
6764172 c.840C>T p.G280G Chr.11: 66668864-66668864 on Build GRCh38
5792340 c.691G>T p.A231S Chr.11: 66669013-66669013 on Build GRCh38
7334425 c.803T>C p.V268A Chr.11: 66668901-66668901 on Build GRCh38
5549221 c.710C>T p.A237V Chr.11: 66668994-66668994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777996819 Chr.11: 66668903-66668903 on Build GRCh38
rs754773604 Chr.11: 66668989-66668989 on Build GRCh38
rs138212594 Chr.11: 66669017-66669017 on Build GRCh38
rs141045331 Chr.11: 66668945-66668945 on Build GRCh38
rs765336366 Chr.11: 66668897-66668897 on Build GRCh38
rs775384561 Chr.11: 66668901-66668901 on Build GRCh38
rs146310422 Chr.11: 66669003-66669003 on Build GRCh38
rs764283037 Chr.11: 66668975-66668975 on Build GRCh38
rs746480100 Chr.11: 66669000-66669000 on Build GRCh38
rs746641399 Chr.11: 66669060-66669060 on Build GRCh38
rs764372966 Chr.11: 66669024-66669024 on Build GRCh38
rs763194930 Chr.11: 66668908-66668908 on Build GRCh38
rs758074854 Chr.11: 66669050-66669050 on Build GRCh38
rs778581838 Chr.11: 66669043-66669043 on Build GRCh38
rs374593903 Chr.11: 66668966-66668966 on Build GRCh38
rs371878027 Chr.11: 66669012-66669012 on Build GRCh38
rs951406935 Chr.11: 66669026-66669026 on Build GRCh38
rs771633119 Chr.11: 66668943-66668943 on Build GRCh38
rs761181043 Chr.11: 66668951-66668951 on Build GRCh38
rs758456481 Chr.11: 66668902-66668902 on Build GRCh38
rs753789368 Chr.11: 66669037-66669037 on Build GRCh38
rs138950956 Chr.11: 66668984-66668984 on Build GRCh38
rs745858956 Chr.11: 66668912-66668912 on Build GRCh38
rs148731962 Chr.11: 66669030-66669030 on Build GRCh38
rs200334725 Chr.11: 66668993-66668993 on Build GRCh38
rs768836554 Chr.11: 66669057-66669057 on Build GRCh38
rs529466760 Chr.11: 66668976-66668976 on Build GRCh38
rs777008884 Chr.11: 66668961-66668961 on Build GRCh38
rs747809725 Chr.11: 66668932-66668932 on Build GRCh38
rs771651424 Chr.11: 66668997-66668997 on Build GRCh38
rs754925060 Chr.11: 66669042-66669042 on Build GRCh38
rs759769612 Chr.11: 66668964-66668964 on Build GRCh38
rs763091259 Chr.11: 66669020-66669020 on Build GRCh38
rs61741747 Chr.11: 66668999-66668999 on Build GRCh38
rs747808883 Chr.11: 66668994-66668994 on Build GRCh38
rs371760573 Chr.11: 66668913-66668913 on Build GRCh38
rs150237340 Chr.11: 66668980-66668980 on Build GRCh38
rs368007657 Chr.11: 66668969-66668969 on Build GRCh38
rs142896309 Chr.11: 66669019-66669019 on Build GRCh38
rs1053443610 Chr.11: 66668963-66668963 on Build GRCh38
rs574603433 Chr.11: 66668892-66668892 on Build GRCh38
rs768380966 Chr.11: 66668923-66668923 on Build GRCh38
rs780093249 Chr.11: 66668981-66668981 on Build GRCh38
rs778483374 Chr.11: 66668927-66668927 on Build GRCh38
rs771359086 Chr.11: 66668957-66668957 on Build GRCh38
rs780002507 Chr.11: 66668915-66668915 on Build GRCh38
rs748998928 Chr.11: 66668921-66668921 on Build GRCh38
rs562073155 Chr.11: 66668941-66668941 on Build GRCh38
rs760057266 Chr.11: 66669013-66669013 on Build GRCh38
rs748255777 Chr.11: 66668999-66669000 on Build GRCh38
rs772480668 Chr.11: 66668944-66668944 on Build GRCh38
rs374733996 Chr.11: 66669049-66669049 on Build GRCh38
rs146095184 Chr.11: 66669025-66669025 on Build GRCh38
rs201202684 Chr.11: 66669036-66669036 on Build GRCh38

View Full Product Details