Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51969744-51969991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD14B
Entrez Gene ID
84836
Gene Name
abhydrolase domain containing 14B
Gene Aliases
CIB, HEL-S-299
UniGene
Hs.420796
Gene Chromosome Location
Chr.3: 51968510-51974630 on Build GRCh38
Transcripts
NM_001146314.1 NM_001254753.1 NM_032750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269238 c.437C>A p.A146D Chr.3: 51969953-51969953 on Build GRCh38
7107266 c.437A>G p.N146S Chr.3: 51969959-51969959 on Build GRCh38
6269237 c.443C>A p.A148D Chr.3: 51969953-51969953 on Build GRCh38
7107267 c.431A>G p.N144S Chr.3: 51969959-51969959 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759988155 Chr.3: 51969779-51969779 on Build GRCh38
rs760191591 Chr.3: 51969920-51969920 on Build GRCh38
rs767029304 Chr.3: 51969806-51969806 on Build GRCh38
rs760314488 Chr.3: 51969823-51969823 on Build GRCh38
rs4687793 Chr.3: 51969872-51969872 on Build GRCh38
rs765061080 Chr.3: 51969941-51969941 on Build GRCh38
rs919670360 Chr.3: 51969850-51969850 on Build GRCh38
rs758354445 Chr.3: 51969950-51969950 on Build GRCh38
rs772639929 Chr.3: 51969919-51969919 on Build GRCh38
rs747155520 Chr.3: 51969854-51969854 on Build GRCh38
rs539332467 Chr.3: 51969809-51969809 on Build GRCh38
rs893241870 Chr.3: 51969763-51969764 on Build GRCh38
rs753572581 Chr.3: 51969827-51969827 on Build GRCh38
rs775764776 Chr.3: 51969916-51969916 on Build GRCh38
rs752493757 Chr.3: 51969944-51969944 on Build GRCh38
rs778675759 Chr.3: 51969833-51969833 on Build GRCh38
rs758318735 Chr.3: 51969841-51969841 on Build GRCh38
rs1034611029 Chr.3: 51969799-51969799 on Build GRCh38
rs751725326 Chr.3: 51969864-51969865 on Build GRCh38
rs770160234 Chr.3: 51969890-51969890 on Build GRCh38
rs763405149 Chr.3: 51969803-51969803 on Build GRCh38
rs769109040 Chr.3: 51969917-51969917 on Build GRCh38
rs1038305294 Chr.3: 51969945-51969945 on Build GRCh38
rs139689394 Chr.3: 51969954-51969954 on Build GRCh38
rs777899228 Chr.3: 51969842-51969842 on Build GRCh38
rs543155783 Chr.3: 51969881-51969881 on Build GRCh38
rs981664081 Chr.3: 51969787-51969787 on Build GRCh38
rs922715125 Chr.3: 51969766-51969766 on Build GRCh38
rs776091750 Chr.3: 51969798-51969798 on Build GRCh38
rs765966516 Chr.3: 51969926-51969926 on Build GRCh38
rs766022003 Chr.3: 51969825-51969825 on Build GRCh38
rs765608281 Chr.3: 51969781-51969781 on Build GRCh38
rs554454985 Chr.3: 51969835-51969835 on Build GRCh38
rs374413054 Chr.3: 51969828-51969828 on Build GRCh38
rs201128395 Chr.3: 51969855-51969855 on Build GRCh38
rs572006151 Chr.3: 51969767-51969767 on Build GRCh38
rs759152293 Chr.3: 51969939-51969939 on Build GRCh38

View Full Product Details