Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15212013-15212274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
UniGene
Hs.595234
Gene Chromosome Location
Chr.3: 15206226-15252916 on Build GRCh38
Transcripts
NM_014296.2 XM_011533555.2 XM_011533556.2 XM_011533557.2 XM_017006043.1 XM_017006044.1 XM_017006045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451638 c.111A>G p.A37A Chr.3: 15212112-15212112 on Build GRCh38
5398436 c.124T>C p.Y42H Chr.3: 15212125-15212125 on Build GRCh38
382272 c.109G>A p.A37T Chr.3: 15212110-15212110 on Build GRCh38
5994509 c.175G>A p.E59K Chr.3: 15212176-15212176 on Build GRCh38
1040084 c.129T>C p.A43A Chr.3: 15212130-15212130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759459747 Chr.3: 15212137-15212137 on Build GRCh38
rs753969497 Chr.3: 15212073-15212073 on Build GRCh38
rs757244189 Chr.3: 15212083-15212083 on Build GRCh38
rs938663568 Chr.3: 15212159-15212159 on Build GRCh38
rs748864995 Chr.3: 15212117-15212117 on Build GRCh38
rs371561470 Chr.3: 15212101-15212101 on Build GRCh38
rs369177764 Chr.3: 15212243-15212243 on Build GRCh38
rs147308439 Chr.3: 15212062-15212062 on Build GRCh38
rs755258990 Chr.3: 15212218-15212218 on Build GRCh38
rs764545432 Chr.3: 15212175-15212175 on Build GRCh38
rs758889233 Chr.3: 15212091-15212091 on Build GRCh38
rs779603414 Chr.3: 15212085-15212085 on Build GRCh38
rs745322596 Chr.3: 15212237-15212237 on Build GRCh38
rs750942548 Chr.3: 15212087-15212087 on Build GRCh38
rs770400520 Chr.3: 15212126-15212126 on Build GRCh38
rs762001265 Chr.3: 15212196-15212196 on Build GRCh38
rs751977326 Chr.3: 15212211-15212211 on Build GRCh38
rs780563309 Chr.3: 15212099-15212099 on Build GRCh38
rs375747480 Chr.3: 15212092-15212092 on Build GRCh38
rs765392901 Chr.3: 15212202-15212202 on Build GRCh38
rs772015323 Chr.3: 15212242-15212242 on Build GRCh38
rs780651270 Chr.3: 15212210-15212210 on Build GRCh38
rs760334490 Chr.3: 15212170-15212170 on Build GRCh38
rs748874148 Chr.3: 15212223-15212223 on Build GRCh38
rs574868976 Chr.3: 15212220-15212220 on Build GRCh38
rs540304739 Chr.3: 15212230-15212230 on Build GRCh38
rs754344102 Chr.3: 15212192-15212204 on Build GRCh38
rs778397013 Chr.3: 15212235-15212235 on Build GRCh38
rs180855962 Chr.3: 15212065-15212065 on Build GRCh38
rs751035768 Chr.3: 15212207-15212207 on Build GRCh38
rs761271512 Chr.3: 15212175-15212176 on Build GRCh38
rs978835488 Chr.3: 15212191-15212191 on Build GRCh38
rs769501079 Chr.3: 15212110-15212110 on Build GRCh38
rs775365951 Chr.3: 15212163-15212163 on Build GRCh38
rs753975969 Chr.3: 15212188-15212188 on Build GRCh38
rs760953519 Chr.3: 15212066-15212066 on Build GRCh38
rs777242348 Chr.3: 15212113-15212113 on Build GRCh38
rs764161167 Chr.3: 15212069-15212069 on Build GRCh38
rs376932100 Chr.3: 15212138-15212138 on Build GRCh38
rs372827755 Chr.3: 15212059-15212059 on Build GRCh38
rs758981120 Chr.3: 15212209-15212209 on Build GRCh38
rs774324879 Chr.3: 15212135-15212135 on Build GRCh38
rs764404005 Chr.3: 15212176-15212176 on Build GRCh38
rs991739983 Chr.3: 15212076-15212076 on Build GRCh38

View Full Product Details