Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63807497-63807770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX42
Entrez Gene ID
11325
Gene Name
DEAD-box helicase 42
Gene Aliases
DDX42P, RHELP, RNAHP, SF3B8, SF3b125
UniGene
Hs.702010
Gene Chromosome Location
Chr.17: 63773856-63819317 on Build GRCh38
Transcripts
NM_007372.3 NM_203499.2 XM_006721657.1 XM_011524259.2 XM_017024111.1 XM_017024112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741091 c.880A>T p.I294F Chr.17: 63807757-63807757 on Build GRCh38
707716 c.883G>T p.G295C Chr.17: 63807760-63807760 on Build GRCh38
3370852 c.879G>A p.M293I Chr.17: 63807756-63807756 on Build GRCh38
982692 c.874G>T p.D292Y Chr.17: 63807751-63807751 on Build GRCh38
6041626 c.847-6C>A p.? Chr.17: 63807718-63807718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374568330 Chr.17: 63807730-63807730 on Build GRCh38
rs761857248 Chr.17: 63807601-63807601 on Build GRCh38
rs1046036090 Chr.17: 63807595-63807595 on Build GRCh38
rs949342450 Chr.17: 63807590-63807590 on Build GRCh38
rs1040728572 Chr.17: 63807563-63807563 on Build GRCh38
rs776662480 Chr.17: 63807577-63807577 on Build GRCh38
rs543730812 Chr.17: 63807536-63807536 on Build GRCh38
rs370109600 Chr.17: 63807702-63807702 on Build GRCh38
rs940371467 Chr.17: 63807650-63807650 on Build GRCh38
rs770451349 Chr.17: 63807733-63807733 on Build GRCh38
rs1037429381 Chr.17: 63807672-63807672 on Build GRCh38
rs1031906101 Chr.17: 63807645-63807645 on Build GRCh38
rs779410079 Chr.17: 63807696-63807696 on Build GRCh38
rs748473905 Chr.17: 63807729-63807729 on Build GRCh38
rs916457476 Chr.17: 63807541-63807541 on Build GRCh38
rs757943914 Chr.17: 63807678-63807678 on Build GRCh38
rs367915242 Chr.17: 63807538-63807538 on Build GRCh38
rs957871419 Chr.17: 63807661-63807661 on Build GRCh38
rs781629106 Chr.17: 63807710-63807710 on Build GRCh38
rs188133606 Chr.17: 63807706-63807706 on Build GRCh38
rs570122370 Chr.17: 63807524-63807524 on Build GRCh38
rs902211881 Chr.17: 63807572-63807572 on Build GRCh38
rs907573214 Chr.17: 63807617-63807617 on Build GRCh38

View Full Product Details