Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184325760-184326015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G1
Entrez Gene ID
1981
Gene Name
eukaryotic translation initiation factor 4 gamma 1
Gene Aliases
EIF-4G1, EIF4F, EIF4G, EIF4GI, P220, PARK18
UniGene
Hs.433750
Gene Chromosome Location
Chr.3: 184314495-184335358 on Build GRCh38
Transcripts
NM_001194946.1 NM_001194947.1 NM_001291157.1 NM_004953.4 NM_182917.4 NM_198241.2 NM_198242.2 NM_198244.2 NR_003055.1

Gene Symbol
SNORD66
Entrez Gene ID
692107
Gene Name
small nucleolar RNA, C/D box 66
Gene Aliases
HBII-142
UniGene
Gene Chromosome Location
Chr.3: 184325696-184325771 on Build GRCh38
Transcripts
NM_001194946.1 NM_001194947.1 NM_001291157.1 NM_004953.4 NM_182917.4 NM_198241.2 NM_198242.2 NM_198244.2 NR_003055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955316 c.3143A>C p.D1048A Chr.3: 184325872-184325872 on Build GRCh38
3590751 c.3123C>T p.S1041S Chr.3: 184325852-184325852 on Build GRCh38
6309001 c.3151G>A p.G1051S Chr.3: 184325880-184325880 on Build GRCh38
479789 c.3202C>T p.R1068* Chr.3: 184325931-184325931 on Build GRCh38
3590752 c.3135C>T p.P1045P Chr.3: 184325864-184325864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747913600 Chr.3: 184325827-184325827 on Build GRCh38
rs267599714 Chr.3: 184325865-184325865 on Build GRCh38
rs775684766 Chr.3: 184325852-184325852 on Build GRCh38
rs897665762 Chr.3: 184325784-184325784 on Build GRCh38
rs754479355 Chr.3: 184325927-184325927 on Build GRCh38
rs957759018 Chr.3: 184325918-184325918 on Build GRCh38
rs997409578 Chr.3: 184325794-184325794 on Build GRCh38
rs868515495 Chr.3: 184325828-184325828 on Build GRCh38
rs760654524 Chr.3: 184325941-184325941 on Build GRCh38
rs754709304 Chr.3: 184325815-184325815 on Build GRCh38
rs184078840 Chr.3: 184325825-184325825 on Build GRCh38
rs904010765 Chr.3: 184325809-184325809 on Build GRCh38
rs767213032 Chr.3: 184325939-184325939 on Build GRCh38
rs199973292 Chr.3: 184325887-184325887 on Build GRCh38
rs564147627 Chr.3: 184325833-184325833 on Build GRCh38
rs763555450 Chr.3: 184325898-184325898 on Build GRCh38
rs774591956 Chr.3: 184325970-184325970 on Build GRCh38
rs888466147 Chr.3: 184325844-184325844 on Build GRCh38
rs757166144 Chr.3: 184325974-184325974 on Build GRCh38
rs777798559 Chr.3: 184325834-184325834 on Build GRCh38
rs1020373331 Chr.3: 184325964-184325964 on Build GRCh38
rs764023335 Chr.3: 184325958-184325958 on Build GRCh38
rs747077459 Chr.3: 184325853-184325853 on Build GRCh38
rs114615711 Chr.3: 184325848-184325848 on Build GRCh38
rs765645340 Chr.3: 184325871-184325871 on Build GRCh38
rs377248755 Chr.3: 184325796-184325796 on Build GRCh38
rs749309199 Chr.3: 184325842-184325842 on Build GRCh38
rs776839352 Chr.3: 184325859-184325859 on Build GRCh38
rs201567302 Chr.3: 184325854-184325854 on Build GRCh38
rs764011884 Chr.3: 184325955-184325955 on Build GRCh38
rs1015283417 Chr.3: 184325919-184325919 on Build GRCh38
rs755976169 Chr.3: 184325852-184325852 on Build GRCh38
rs751139529 Chr.3: 184325800-184325800 on Build GRCh38
rs778743138 Chr.3: 184325983-184325983 on Build GRCh38
rs762105656 Chr.3: 184325864-184325864 on Build GRCh38
rs747042053 Chr.3: 184325985-184325985 on Build GRCh38
rs368921147 Chr.3: 184325792-184325792 on Build GRCh38
rs143983618 Chr.3: 184325903-184325903 on Build GRCh38
rs765785554 Chr.3: 184325899-184325899 on Build GRCh38
rs146433145 Chr.3: 184325947-184325947 on Build GRCh38
rs757830669 Chr.3: 184325791-184325791 on Build GRCh38
rs373591739 Chr.3: 184325913-184325913 on Build GRCh38
rs765980019 Chr.3: 184325799-184325799 on Build GRCh38
rs1031980532 Chr.3: 184325855-184325855 on Build GRCh38

View Full Product Details