Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 145288-145539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB126
Entrez Gene ID
81623
Gene Name
defensin beta 126
Gene Aliases
C20orf8, DEFB-26, DEFB26, HBD26, bA530N10.1, hBD-26
UniGene
Hs.124211
Gene Chromosome Location
Chr.20: 142553-145751 on Build GRCh38
Transcripts
NM_030931.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096652 c.59-2A>G p.? Chr.20: 145413-145413 on Build GRCh38
1661431 c.161A>G p.K54R Chr.20: 145517-145517 on Build GRCh38
5689213 c.77A>T p.K26M Chr.20: 145433-145433 on Build GRCh38
3963308 c.149C>A p.A50E Chr.20: 145505-145505 on Build GRCh38
7224607 c.149C>T p.A50V Chr.20: 145505-145505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962468838 Chr.20: 145399-145399 on Build GRCh38
rs190798121 Chr.20: 145475-145475 on Build GRCh38
rs916505927 Chr.20: 145452-145452 on Build GRCh38
rs772800263 Chr.20: 145512-145512 on Build GRCh38
rs901620115 Chr.20: 145518-145519 on Build GRCh38
rs201416351 Chr.20: 145513-145513 on Build GRCh38
rs1056376458 Chr.20: 145451-145451 on Build GRCh38
rs772082992 Chr.20: 145447-145447 on Build GRCh38
rs751216582 Chr.20: 145390-145390 on Build GRCh38
rs745824481 Chr.20: 145463-145463 on Build GRCh38
rs767029386 Chr.20: 145396-145396 on Build GRCh38
rs762685253 Chr.20: 145383-145383 on Build GRCh38
rs11467497 Chr.20: 145515-145518 on Build GRCh38
rs200901241 Chr.20: 145520-145520 on Build GRCh38
rs769611855 Chr.20: 145364-145364 on Build GRCh38
rs200807952 Chr.20: 145514-145517 on Build GRCh38
rs763583575 Chr.20: 145388-145388 on Build GRCh38
rs768487465 Chr.20: 145470-145470 on Build GRCh38
rs751009497 Chr.20: 145408-145408 on Build GRCh38
rs75442118 Chr.20: 145508-145508 on Build GRCh38
rs766966497 Chr.20: 145443-145443 on Build GRCh38
rs754383543 Chr.20: 145518-145518 on Build GRCh38
rs755346738 Chr.20: 145519-145519 on Build GRCh38
rs759127488 Chr.20: 145436-145436 on Build GRCh38
rs768976288 Chr.20: 145513-145516 on Build GRCh38
rs769657189 Chr.20: 145465-145465 on Build GRCh38
rs182093028 Chr.20: 145494-145494 on Build GRCh38
rs532491188 Chr.20: 145458-145458 on Build GRCh38
rs775273263 Chr.20: 145468-145468 on Build GRCh38
rs749108147 Chr.20: 145469-145469 on Build GRCh38
rs397788846 Chr.20: 145519-145522 on Build GRCh38
rs575457625 Chr.20: 145336-145336 on Build GRCh38
rs561390827 Chr.20: 145423-145423 on Build GRCh38
rs184470868 Chr.20: 145434-145434 on Build GRCh38
rs771000519 Chr.20: 145354-145355 on Build GRCh38
rs200600942 Chr.20: 145501-145501 on Build GRCh38
rs749729924 Chr.20: 145459-145461 on Build GRCh38
rs972948427 Chr.20: 145431-145431 on Build GRCh38
rs142181217 Chr.20: 145367-145367 on Build GRCh38
rs112386635 Chr.20: 145509-145509 on Build GRCh38
rs146358503 Chr.20: 145444-145444 on Build GRCh38
rs754197683 Chr.20: 145438-145438 on Build GRCh38
rs930567514 Chr.20: 145454-145454 on Build GRCh38
rs780271265 Chr.20: 145450-145450 on Build GRCh38
rs142956939 Chr.20: 145459-145459 on Build GRCh38
rs34651560 Chr.20: 145446-145446 on Build GRCh38
rs773854587 Chr.20: 145333-145333 on Build GRCh38
rs867653623 Chr.20: 145518-145519 on Build GRCh38
rs960983335 Chr.20: 145413-145413 on Build GRCh38
rs761284364 Chr.20: 145393-145393 on Build GRCh38

View Full Product Details