Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232870239-232870508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf82
Entrez Gene ID
389084
Gene Name
chromosome 2 open reading frame 82
Gene Aliases
UNQ830
UniGene
Hs.657509
Gene Chromosome Location
Chr.2: 232865678-232878700 on Build GRCh38
Transcripts
NM_206895.1 XM_017004080.1 XM_017004081.1 XM_017004082.1 XM_017004083.1 XM_017004084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1406304 c.63G>A p.A21A Chr.2: 232870404-232870404 on Build GRCh38
1406303 c.63G>T p.A21A Chr.2: 232870404-232870404 on Build GRCh38
1198744 c.23G>A p.R8H Chr.2: 232870364-232870364 on Build GRCh38
5951058 c.29C>T p.A10V Chr.2: 232870370-232870370 on Build GRCh38
7199374 c.44C>T p.S15F Chr.2: 232870385-232870385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916180950 Chr.2: 232870282-232870282 on Build GRCh38
rs760213130 Chr.2: 232870334-232870334 on Build GRCh38
rs775289630 Chr.2: 232870429-232870429 on Build GRCh38
rs756152920 Chr.2: 232870370-232870370 on Build GRCh38
rs771759126 Chr.2: 232870326-232870329 on Build GRCh38
rs779322861 Chr.2: 232870404-232870404 on Build GRCh38
rs372317872 Chr.2: 232870313-232870313 on Build GRCh38
rs371543923 Chr.2: 232870333-232870333 on Build GRCh38
rs374593527 Chr.2: 232870478-232870478 on Build GRCh38
rs948765190 Chr.2: 232870270-232870270 on Build GRCh38
rs114532944 Chr.2: 232870452-232870452 on Build GRCh38
rs779387952 Chr.2: 232870386-232870386 on Build GRCh38
rs35775435 Chr.2: 232870339-232870340 on Build GRCh38
rs776561447 Chr.2: 232870349-232870349 on Build GRCh38
rs371963662 Chr.2: 232870403-232870403 on Build GRCh38
rs369644446 Chr.2: 232870364-232870364 on Build GRCh38
rs750537320 Chr.2: 232870369-232870369 on Build GRCh38
rs751721233 Chr.2: 232870295-232870295 on Build GRCh38
rs530709180 Chr.2: 232870358-232870358 on Build GRCh38
rs201000279 Chr.2: 232870328-232870328 on Build GRCh38
rs530304013 Chr.2: 232870294-232870294 on Build GRCh38
rs138245588 Chr.2: 232870398-232870398 on Build GRCh38
rs750586693 Chr.2: 232870460-232870460 on Build GRCh38
rs200597442 Chr.2: 232870360-232870360 on Build GRCh38
rs764992998 Chr.2: 232870423-232870423 on Build GRCh38
rs757383182 Chr.2: 232870296-232870296 on Build GRCh38
rs367756993 Chr.2: 232870330-232870330 on Build GRCh38
rs772959887 Chr.2: 232870394-232870394 on Build GRCh38
rs773934126 Chr.2: 232870319-232870319 on Build GRCh38
rs868376319 Chr.2: 232870480-232870480 on Build GRCh38
rs199967566 Chr.2: 232870363-232870363 on Build GRCh38
rs373689575 Chr.2: 232870317-232870317 on Build GRCh38
rs766477716 Chr.2: 232870464-232870464 on Build GRCh38
rs1046920221 Chr.2: 232870307-232870307 on Build GRCh38
rs747850236 Chr.2: 232870387-232870387 on Build GRCh38
rs199594735 Chr.2: 232870371-232870371 on Build GRCh38
rs917238433 Chr.2: 232870260-232870260 on Build GRCh38
rs1024407034 Chr.2: 232870444-232870444 on Build GRCh38
rs920648091 Chr.2: 232870301-232870301 on Build GRCh38
rs968490175 Chr.2: 232870338-232870338 on Build GRCh38
rs775268968 Chr.2: 232870370-232870371 on Build GRCh38
rs776487003 Chr.2: 232870413-232870413 on Build GRCh38
rs751823425 Chr.2: 232870361-232870361 on Build GRCh38
rs749619568 Chr.2: 232870316-232870316 on Build GRCh38
rs916128438 Chr.2: 232870399-232870399 on Build GRCh38
rs755367305 Chr.2: 232870385-232870385 on Build GRCh38
rs992943752 Chr.2: 232870258-232870258 on Build GRCh38
rs781221390 Chr.2: 232870303-232870303 on Build GRCh38
rs201190318 Chr.2: 232870308-232870308 on Build GRCh38
rs542263903 Chr.2: 232870299-232870299 on Build GRCh38
rs369842484 Chr.2: 232870462-232870462 on Build GRCh38
rs766255599 Chr.2: 232870335-232870335 on Build GRCh38
rs749791125 Chr.2: 232870372-232870372 on Build GRCh38
rs762699423 Chr.2: 232870447-232870447 on Build GRCh38
rs757771157 Chr.2: 232870390-232870390 on Build GRCh38
rs759510202 Chr.2: 232870422-232870422 on Build GRCh38

View Full Product Details