Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43399829-43400080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL4D
Entrez Gene ID
379
Gene Name
ADP ribosylation factor like GTPase 4D
Gene Aliases
ARF4L, ARL6
UniGene
Hs.183153
Gene Chromosome Location
Chr.17: 43398985-43401136 on Build GRCh38
Transcripts
NM_001661.3 XM_011524782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2155758 c.136T>A p.F46I Chr.17: 43399868-43399868 on Build GRCh38
2155757 c.128T>C p.F43S Chr.17: 43399860-43399860 on Build GRCh38
5689125 c.133G>A p.E45K Chr.17: 43399865-43399865 on Build GRCh38
2155756 c.129C>G p.F43L Chr.17: 43399861-43399861 on Build GRCh38
5578064 c.341T>C p.V114A Chr.17: 43400073-43400073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758999530 Chr.17: 43399919-43399919 on Build GRCh38
rs955391996 Chr.17: 43400043-43400043 on Build GRCh38
rs753313438 Chr.17: 43399945-43399945 on Build GRCh38
rs765651568 Chr.17: 43400055-43400055 on Build GRCh38
rs760421620 Chr.17: 43400053-43400053 on Build GRCh38
rs775275422 Chr.17: 43399963-43399963 on Build GRCh38
rs761216623 Chr.17: 43400042-43400042 on Build GRCh38
rs781253273 Chr.17: 43399952-43399952 on Build GRCh38
rs758198899 Chr.17: 43399963-43399964 on Build GRCh38
rs779429695 Chr.17: 43399913-43399913 on Build GRCh38
rs750033537 Chr.17: 43400051-43400051 on Build GRCh38
rs769510993 Chr.17: 43399961-43399961 on Build GRCh38
rs149623988 Chr.17: 43399939-43399939 on Build GRCh38
rs767054676 Chr.17: 43399851-43399851 on Build GRCh38
rs774397233 Chr.17: 43400017-43400017 on Build GRCh38
rs1059968 Chr.17: 43400004-43400004 on Build GRCh38
rs11545626 Chr.17: 43399997-43399997 on Build GRCh38
rs147319702 Chr.17: 43399960-43399960 on Build GRCh38
rs753200298 Chr.17: 43399914-43399914 on Build GRCh38
rs371969086 Chr.17: 43399864-43399864 on Build GRCh38
rs768448857 Chr.17: 43399973-43399973 on Build GRCh38
rs767123284 Chr.17: 43400047-43400047 on Build GRCh38
rs935334904 Chr.17: 43400010-43400010 on Build GRCh38
rs750148648 Chr.17: 43399853-43399853 on Build GRCh38
rs924018125 Chr.17: 43399995-43399995 on Build GRCh38
rs368069774 Chr.17: 43400044-43400044 on Build GRCh38
rs755394263 Chr.17: 43399870-43399870 on Build GRCh38
rs148824656 Chr.17: 43399969-43399969 on Build GRCh38
rs779395257 Chr.17: 43399924-43399924 on Build GRCh38

View Full Product Details